PloS one
A Role of Myocardin Related Transcription Factor-A (MRTF-A) in Scleroderma Related Fibrosis.
PMID 25955164
Abstract
In scleroderma (systemic sclerosis, SSc), persistent activation of myofibroblast leads to severe skin and organ fibrosis resistant to therapy. Increased mechanical stiffness in the involved fibrotic tissues is a hallmark clinical feature and a cause of disabling symptoms. Myocardin Related Transcription Factor-A (MRTF-A) is a transcriptional co-activator that is sequestered in the cytoplasm and translocates to the nucleus under mechanical stress or growth factor stimulation. Our objective was to determine if MRTF-A is activated in the disease microenvironment to produce more extracellular matrix in progressive SSc. Immunohistochemistry studies demonstrate that nuclear translocation of MRTF-A in scleroderma tissues occurs in keratinocytes, endothelial cells, infiltrating inflammatory cells, and dermal fibroblasts, consistent with enhanced signaling in multiple cell lineages exposed to the stiff extracellular matrix. Inhibition of MRTF-A nuclear translocation or knockdown of MRTF-A synthesis abolishes the SSc myofibroblast enhanced basal contractility and synthesis of type I collagen and inhibits the matricellular profibrotic protein, connective tissue growth factor (CCN2/CTGF). In MRTF-A null mice, basal skin and lung stiffness was abnormally reduced and associated with altered fibrillar collagen. MRTF-A has a role in SSc fibrosis acting as a central regulator linking mechanical cues to adverse remodeling of the extracellular matrix.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4871
Anti-CTGF (181-195) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1300909
Anti-CTGF (C-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1300910
Anti-CTGF (center) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
HPA031074
Anti-CTGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA031075
Anti-CTGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
AV37504
Anti-MKL1 antibody produced in rabbit, IgG fraction of antiserum
HPA030782
Anti-MKL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
EHU015721 MISSION® esiRNA, esiRNA targeting human MKL1 (esiRNA1)
EMU073461 MISSION® esiRNA, esiRNA targeting mouse Mkl1 (esiRNA1)
SAB4200401
Monoclonal Anti-CTGF antibody produced in mouse, ~1.0 mg/mL, clone CTGF-14, purified immunoglobulin
SAB1403710 Monoclonal Anti-CTGF antibody produced in mouse, clone 4D9, purified immunoglobulin, buffered aqueous solution
SAB1402162
Monoclonal Anti-CTGF antibody produced in mouse, clone 2C11, purified immunoglobulin, buffered aqueous solution