EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
Oncogenic Activity of miR-650 in Prostate Cancer Is Mediated by Suppression of CSR1 Expression.
PMID 25956032
Abstract
Cellular stress response 1 (CSR1) is a tumor suppressor gene whose expression was frequently down-regulated in prostate cancer. The mechanism of its down-regulation, however, is not clear. Here, we show that the 3' untranslated region of CSR1 contains a target site of miR-650. High level of miR-650 was found in prostate cancer samples and cell lines. Degradation of miR-650 by specific inhibitor dramatically increased the expression levels of CSR1. Interaction between miR-650 and its target site in the 3' untranslated region was validated through luciferase reporter system. Mutation at the target site completely abrogated the activity of miR-650 on the 3' untranslated region of CSR1. Inhibition of miR-650 reversed the expression suppression of CSR1, suppressed colony formation, and blocked cell cycle entry to the S phase of both PC3 and DU145 cells. Animal model showed significant decrease of tumor volume, rate of metastasis, and mortality of severe combined immunodeficient mice xenografted with PC3 or DU145 cells transformed with inhibitor of miR-650. Our analyses demonstrate that suppression of CSR1 expression is a novel mechanism critical for the oncogenic activity of miR-650.