EMAIL THIS PAGE TO A FRIEND
Cellular signalling
Cooperation of distinct Rac-dependent pathways to stabilise E-cadherin adhesion.
PMID 25957131
Abstract
The precise mechanisms via which Rac1 is activated by cadherin junctions are not fully known. In keratinocytes Rac1 activation by cadherin junctions requires EGFR signalling, but how EGFR does so is unclear. To address which activator could mediate E-cadherin signalling to Rac1, we investigated EGFR and two Rac1 GEFs, SOS1 and DOCK180. EGFR RNAi prevented junction-induced Rac1 activation and led to fragmented localization of E-cadherin at cadherin contacts. In contrast, depletion of another EGFR family member, ErbB3, did not interfere with either process. DOCK180 RNAi, but not SOS1, prevented E-cadherin-induced Rac1 activation. However, in a strong divergence from EGFR RNAi phenotype, DOCK180 depletion did not perturb actin recruitment or cadherin localisation at junctions. Rather, reduced DOCK180 levels impaired the resistance to mechanical stress of pre-formed cell aggregates. Thus, within the same cell type, EGFR and DOCK180 regulate Rac1 activation by newly-formed contacts, but control separate cellular events that cooperate to stabilise junctions.