EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Immobilization of nitrate reductase onto epoxy affixed silver nanoparticles for determination of soil nitrates.
PMID 25957718
Abstract
Epoxy glued silver nanoparticles were used as immobilization support for nitrate reductase (NR). The resulting epoxy/AgNPs/NR conjugates were characterized at successive stages of fabrication by scanning electron microscopy and fourier transform infrared spectroscopy. The immobilized enzyme system exhibited reasonably high conjugation yield (37.6±0.01 μg/cm(2)), with 93.54±0.88% retention of specific activity. Most favorable working conditions of pH, temperature and substrate concentration were ascertained to optimize the performance of epoxy/AgNPs/NR conjugates for soil nitrate quantification. The analytical results for soil nitrate determination were consistent, reliable and reproducible. Minimum detection limit of the method was 0.05 mM with linearity from 0.1 to 11.0 mM. The % recoveries of added nitrates (0.1 and 0.2 mM) were<95.0% and within-day and between-day coefficients of variations were 0.556% and 1.63% respectively. The method showed good correlation (R(2)=0.998) with the popular Griess reaction method. Epoxy/AgNPs bound NR had a half-life of 18 days at 4 °C and retained 50% activity after 15 reuses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
674427
11-Mercaptoundecanoic acid, 98%
C11H22O2S
450561
11-Mercaptoundecanoic acid, 95%
C11H22O2S