EMAIL THIS PAGE TO A FRIEND
FEBS letters
Clec4g (LSECtin) interacts with BACE1 and suppresses Aβ generation.
PMID 25957769
Abstract
β-Site amyloid precursor protein cleaving enzyme-1 (BACE1) is a central molecule in Alzheimer's disease (AD). It cleaves amyloid precursor protein (APP) to produce the toxic amyloid-β (Aβ) peptides. Thus, a novel BACE1 modulator could offer a new therapeutic strategy for AD. We report that C-type lectin-like domain family 4, member g (Clec4g, also designated as LSECtin) interacts with BACE1 in mouse brain and cultured cells. Overexpression of Clec4g suppressed BACE1-mediated Aβ generation, and affected the intracellular distribution of BACE1 but not its catalytic activity. These results highlight a novel role of Clec4g in negatively regulating BACE1 function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500240
Anti-BACE antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4501017
Anti-BACE, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2900193
Anti-BACE1 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900194
Anti-BACE1 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900206
Anti-BACE1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100200
Anti-BACE1 antibody produced in rabbit, affinity isolated antibody
HPA014044
Anti-CLEC4G antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1304456
ANTI-PDIA2 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2501845
Anti-PDIA2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501851
Anti-PDIA2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA051692
Anti-PDIA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA053492
Anti-PDIA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405897
Anti-PDIA3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002645
Anti-PDIA3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107799
Anti-PDIA3 antibody produced in rabbit, affinity isolated antibody
SAB1410582
Anti-PDIA3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1404579
Monoclonal Anti-EEA1, (C-terminal) antibody produced in mouse, clone 2G2, purified immunoglobulin, buffered aqueous solution