EMAIL THIS PAGE TO A FRIEND
Molecular oncology
Diphtheria-toxin based anti-human CCR4 immunotoxin for targeting human CCR4(+) cells inxa0vivo.
PMID 25958791
Abstract
CC chemokine receptor 4 (CCR4) has attracted much attention as a promising therapeutic drug target for CCR4(+) tumor cells and Tregs. CCR4 is expressed on some tumor cells such as T-cell acute lymphoblastic leukemia (ALL), adult T-cell leukemia/lymphoma (ATLL), adult peripheral T cell lymphoma (PTCL) and cutaneous T cell lymphoma (CTCL). CCR4 is also expressed on majority of Tregs, mainly effector Tregs. In this study we have successfully developed three versions of diphtheria-toxin based anti-human CCR4 immunotoxins (monovalent, bivalent and single-chain fold-back diabody). Binding analysis by flow cytometry showed that all three versions of the anti-human CCR4 immunotoxins bound to the human CCR4(+) tumor cell line as well as CCR4(+) human PBMC. The bivalent isoform bound stronger than its monovalent counterpart and the single-chain foldback diabody isoform was the strongest among the three versions. Inxa0vitro efficacy analysis demonstrated that the bivalent isoform was 20 fold more potent in inhibiting cellular proliferation and protein synthesis in human CCR4(+) tumor cells compared to the monovalent anti-human CCR4 immunotoxin. The single-chain fold-back diabody isoform was 10 fold more potent than its bivalent counterpart and 200 fold more potent than its monovalent counterpart. The inxa0vivo efficacy was assessed using a human CCR4(+) tumor-bearing mouse model. The immunotoxin significantly prolonged the survival of tumor-bearing NOD/SCID IL-2 receptor γ(-/-) (NSG) mice injected with human CCR4(+) acute lymphoblastic leukemia cells compared with the control group. This novel anti-human CCR4 immunotoxin is a promising drug candidate for targeting human CCR4(+) tumor cells and Tregs inxa0vivo.