EMAIL THIS PAGE TO A FRIEND
Neurochemistry international
Wnt co-receptor LRP5/6 overexpression confers protection against hydrogen peroxide-induced neurotoxicity and reduces tau phosphorylation in SH-SY5Y cells.
PMID 25959626
Abstract
Emerging studies have suggested the involvement of dysregulated Wnt/β-catenin cascade in the etiology of Alzheimer's disease (AD). Recently, genetic variations in Wnt co-receptor low density lipoprotein receptor-related protein (LRP) 6 causing reduced Wnt signaling has been linked to late-onset AD. Here, we hypothesized that overexpression of Wnt co-receptors LRP5 and LRP6 would serve as an effective new approach in reducing neurotoxicity induced by oxidative stress and decreasing tau phosphorylation in SH-SY5Y human neuroblastoma cells. Our results showed that overexpression of LRP5 and LRP6 in SH-SY5Y cells activates Wnt signaling and downstream proliferation genes, whereas knockdown of the co-receptors represses Wnt signaling and the transcription of proliferative markers. We further demonstrated that overexpression of LRP5 and LRP6 protects SH-SY5Y from cell death caused by hydrogen peroxide-induced oxidative stress, inhibits GSK3β activity and subsequently reduces tau phosphorylation. Together, our findings suggest that rescuing LRP5/6-mediated Wnt signaling improves neuronal cell survival and reduces tau phosphorylation, which support the hypothesis that Wnt signaling might be an attractive therapeutic strategy for managing AD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU081381 MISSION® esiRNA, esiRNA targeting mouse Lrp5 (esiRNA1)