EMAIL THIS PAGE TO A FRIEND
Muscle & nerve
Intermittent stretching induces fibrosis in denervated rat muscle.
PMID 25960249
Abstract
Stretching (St) has been used for treating denervated muscles. However, its effectiveness and safety claims require further study. Rats were divided into: (1) those with denervated (D) muscles, evaluated 7 or 15 days after sciatic nerve crush injury; (2) those with D muscles submitted to St during 7 or 15 days; and (3) those with normal muscles. Muscle fiber cross-sectional area, serial sarcomere number, sarcomere length, and connective tissue density were measured. MMP-2, MMP-9, TIMP-1, TGF-β1, and myostatin mRNAs were determined by real-time polymerase chain reaction. MMP-2 and MMP-9 activity was evaluated by zymography. Collagen I was localized using immunofluorescence. St did not prevent muscle atrophy due to denervation, but it increased fibrosis and collagen I deposition at day 15. St also upregulated MMP-9 and TGF-β1 gene expressions at day 7, and myostatin at day 15. Stretching denervated muscle does not prevent atrophy, but it increases fibrosis via temporal modulation of TGF-β1/myostatin and MMP-9 cascades.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6148
Paraformaldehyde, reagent grade, crystalline
CH2O ]n