EMAIL THIS PAGE TO A FRIEND
Scientific reports
Larynx carcinoma regulates tumor-associated macrophages through PLGF signaling.
PMID 25961789
Abstract
Cancer neovascularization plays an essential role in the metastasis of larynx carcinoma (LC). However, the underlying molecular mechanisms are not completely understood. Recently, we reported that placental growth factor (PLGF) regulates expression of matrix metalloproteinase 3 (MMP3) through ERK/MAPK signaling pathway in LC. Here, we show that MMP9 upregulated in LC, and appeared to be mainly produced by M2 macrophages (tumor-associated macrophages (TAM)). In a transwell co-culture system, PLGF secreted by LC cells triggered macrophage polarization to a TAM subtype that releases MMP9. Moreover, MMP9 was found to be activated in the PLGF-polarized TAM via transforming growth factor β (TGFβ) receptor signaling activation. Furthermore, PLGF in LC cells induced macrophage polarization in vivo, and significantly promoted the growth of LC. Thus, together with our previous work, our study highlights a pivotal role of cross-talk between TAM and LC in regulating the metastasis of LC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405819
Anti-FLT1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA011740
Anti-FLT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
SAB1408577
Anti-FLT1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300564
Anti-SMAD3 (Ab-208) antibody produced in rabbit, affinity isolated antibody
SAB4300565
Anti-SMAD3 (Ab-425) antibody produced in rabbit, affinity isolated antibody
AV100621
Anti-SMAD3 antibody produced in rabbit, IgG fraction of antiserum
SAB4200342
Anti-SMAD3 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB2702052
Anti-SMAD3 antibody produced in rabbit
SAB4501811
Anti-SMAD3 antibody produced in rabbit, affinity isolated antibody
SAB4501812
Anti-SMAD3 antibody produced in rabbit, affinity isolated antibody
SAB4501813
Anti-SMAD3 antibody produced in rabbit, affinity isolated antibody
SAB4501810
Anti-SMAD3, C-Terminal antibody produced in rabbit, affinity isolated antibody
V1139 Anti-Vascular Endothelial Growth Factor Receptor 1 antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4500957
Anti-VEGFR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500958
Anti-VEGFR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
RAB0372
Human MMP-9 ELISA Kit, for serum, plasma, cell culture supernatant and urine
EMU183631 MISSION® esiRNA, esiRNA targeting mouse Mki67
AMAB90703
Monoclonal Anti-FLT1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0344, purified immunoglobulin, buffered aqueous glycerol solution
WH0004088M1
Monoclonal Anti-SMAD3 antibody produced in mouse, clone 2C12, purified immunoglobulin, buffered aqueous solution
WH0004088M5 Monoclonal Anti-SMAD3 antibody produced in mouse, clone 4D5, purified immunoglobulin, buffered aqueous solution
SAB1400160
Monoclonal Anti-SMAD3 antibody produced in mouse, clone 7F3, purified immunoglobulin, buffered aqueous solution
SAB1404037
Monoclonal Anti-SMAD3 antibody produced in mouse, clone 4F9, purified immunoglobulin, buffered aqueous solution
V4762 Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-1 antibody produced in mouse, clone FLT-19, tissue culture supernatant
V4262
Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-1 antibody produced in mouse, clone FLT-11, ascites fluid
RAB0373
Mouse pro-MMP-9 ELISA Kit, for serum, plasma and cell culture supernatant
P1588 Placenta Growth Factor human, PlGF, recombinant, expressed in E. coli, powder, suitable for cell culture
SRP4743 PLGF from mouse, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)