EMAIL THIS PAGE TO A FRIEND
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society
Up-Regulation of miR-204 Enhances Anoikis Sensitivity in Epithelial Ovarian Cancer Cell Line Via Brain-Derived Neurotrophic Factor Pathway In Vitro.
PMID 25962115
Abstract
Genomic loci encoding miR-204, which was predicted to target brain-derived neurotrophic factor (BDNF), were frequently lost in multiple cancer, including epithelial ovarian cancer (EOC). In this study, we aimed to find out the influence of miR-204 expression level on EOC cell anoikis sensitivity and to explore possible mechanisms of this process. First, we screened EOC cells, which maintain anoikis resistance forming an anoikis pattern. miR-204 expression level and apoptosis were measured, respectively, by quantitative reverse transcriptase polymerase chain reaction and Annexin-V-R-PE/7-amino-actinomycin assay. Then we restored the expression level of miR-204 by transfection with pre-miR-204. miR-204 expression level and apoptosis were measured as before; cell invasion and migration ability were detected by transwell invasion assay and wound-healing assay. The messenger RNA level of BDNF was also detected by quantitative reverse transcriptase polymerase chain reaction; Western blot analysis was performed to assess pAKT expression. Expression of miR-204 is significantly down-regulated in an anoikis pattern. Restored expression level of miR-204 enables cells to acquire more sensitivity to anoikis and decrease invasive and metastatic behavior, and also results in BDNF down-expression and inhibits activation of mitochondria-dependent pathway through the PI3K/AKT signaling pathway leading to cancer cell anoikis in EOC cells. miR-204 up-regulation may be linked directly to the sensitivity of EOC cell anoikis by contributing to BDNF down-regulation. Our findings provide a novel mechanism for manipulating miR-204 levels therapeutically to restore anoikis sensitivity.