PloS one
Branchial cilia and sperm flagella recruit distinct axonemal components.
PMID 25962172
Abstract
Eukaryotic cilia and flagella have highly conserved 9 + 2 structures. They are functionally diverged to play cell-type-specific roles even in a multicellular organism. Although their structural components are therefore believed to be common, few studies have investigated the molecular diversity of the protein components of the cilia and flagella in a single organism. Here we carried out a proteomic analysis and compared protein components between branchial cilia and sperm flagella in a marine invertebrate chordate, Ciona intestinalis. Distinct feature of protein recruitment in branchial cilia and sperm flagella has been clarified; (1) Isoforms of α- and β-tubulins as well as those of actins are distinctly used in branchial cilia or sperm flagella. (2) Structural components, such as dynein docking complex, tektins and an outer dense fiber protein, are used differently by the cilia and flagella. (3) Sperm flagella are specialized for the cAMP- and Ca2+-dependent regulation of outer arm dynein and for energy metabolism by glycolytic enzymes. Our present study clearly demonstrates that flagellar or ciliary proteins are properly recruited according to their function and stability, despite their apparent structural resemblance and conservation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6528
Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
T7782 Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701031 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701028 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701011 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701033 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Texas Red™ antibody produced in goat, affinity isolated antibody, lyophilized powder
B7401 Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B9904 Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B7022 Anti-Mouse IgG (γ-chain specific)−Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4600013
Anti-Mouse IgG (H+L), CF 350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600019
Anti-Mouse IgG (H+L), CF 405M antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600023
Anti-Mouse IgG (H+L), CF 405S antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600042
Anti-Mouse IgG (H+L), CF 488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600066
Anti-Mouse IgG (H+L), CF 555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600082
Anti-Mouse IgG (H+L), CF 568 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600105
Anti-Mouse IgG (H+L), CF 594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600138
Anti-Mouse IgG (H+L), CF 633 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600161
Anti-Mouse IgG (H+L), CF 640R antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600182
Anti-Mouse IgG (H+L), CF 647 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600191
Anti-Mouse IgG (H+L), CF 660C antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600194
Anti-Mouse IgG (H+L), CF 660R antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600211
Anti-Mouse IgG (H+L), CF 750 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600222
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600405
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600387
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600388
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600299
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600390
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600401
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF568 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600321
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution