EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Hydrophobic-hydrophilic monolithic dual-phase layer for two-dimensional thin-layer chromatography coupled with surface-enhanced Raman spectroscopy detection.
PMID 25962376
Abstract
Hydrophobic-hydrophilic monolithic dual-phase plates have been prepared by a two-step polymerization method for two-dimensional thin-layer chromatography of low-molecular-weight compounds, namely, several dyes. The thin 200 μm poly(glycidyl methacrylate-co-ethylene dimethacrylate) layers attached to microscope glass plates were prepared using a UV-initiated polymerization method within a simple glass mold. After cutting and cleaning the specific area of the layer, the reassembled mold was filled with a polymerization mixture of butyl methacrylate and ethylene dimethacrylate and subsequently irradiated with UV light. During the second polymerization process, the former layer was protected from the UV light with a UV mask. After extracting the porogens and hydrolyzing the poly(glycidyl methacrylate-co-ethylene dimethacrylate) area, these two-dimensional layers were used to separate a mixture of dyes with great difference in their polarity using reversed-phase chromatography mode within the hydrophobic layer and then hydrophilic interaction chromatography mode along the hydrophilic area. In the latter dimension only the specific spot was developed further. Detection of the separated dyes could be achieved with surface-enhanced Raman spectroscopy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
250198
Methyl Red, ACS reagent, crystalline
C15H15N3O2