EMAIL THIS PAGE TO A FRIEND
Infection and immunity
Interaction of Salmonella enterica Serovar Typhimurium with Intestinal Organoids Derived from Human Induced Pluripotent Stem Cells.
PMID 25964470
Abstract
The intestinal mucosa forms the first line of defense against infections mediated by enteric pathogens such as salmonellae. Here we exploited intestinal "organoids" (iHOs) generated from human induced pluripotent stem cells (hIPSCs) to explore the interaction of Salmonella enterica serovar Typhimurium with iHOs. Imaging and RNA sequencing were used to analyze these interactions, and clear changes in transcriptional signatures were detected, including altered patterns of cytokine expression after the exposure of iHOs to bacteria. S. Typhimurium microinjected into the lumen of iHOs was able to invade the epithelial barrier, with many bacteria residing within Salmonella-containing vacuoles. An S. Typhimurium invA mutant defective in the Salmonella pathogenicity island 1 invasion apparatus was less capable of invading the iHO epithelium. Hence, we provide evidence that hIPSC-derived organoids are a promising model of the intestinal epithelium for assessing interactions with enteric pathogens.