EMAIL THIS PAGE TO A FRIEND
Anticancer research
Demethoxycurcumin-induced DNA Damage Decreases DNA Repair-associated Protein Expression Levels in NCI-H460 Human Lung Cancer Cells.
PMID 25964547
Abstract
Demethoxycurcumin (DMC) is a key component of Chinese medicine (Turmeric) and has been proven effective in killing various cancer cells. Its role in inducing cytotoxic effects in many cancer cells has been reported, but its role regarding DNA damage on lung cancer cells has not been studied in detail. In the present study, we demonstrated DMC-induced DNA damage and condensation in NCI-H460 cells by using the Comet assay and DAPI staining examinations, respectively. Western blotting indicated that DMC suppressed the protein levels associated with DNA damage and repair, such as 14-3-3σ (an important checkpoint keeper of DNA damage response), DNA repair proteins breast cancer 1, early onset (BRCA1), O6-methylguanine-DNA methyltransferase (MGMT), mediator of DNA damage checkpoint 1 (MDC1), and p53 (tumor suppressor protein). DMC activated phosphorylated p53 and p-H2A.X (phospho Ser140) in NCI-H460 cells. Furthermore, we used confocal laser systems microscopy to examine the protein translocation. The results showed that DMC promotes the translocation of p-p53 and p-H2A.X from the cytosol to the nuclei in NCI-H460 cells. Taken together, DMC induced DNA damage and affected DNA repair proteins in NCI-H460 cells in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21975F Anti-MGMT antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1406122
Anti-MGMT antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2105451
Anti-MGMT antibody produced in rabbit, affinity isolated antibody
SAB4504499
Anti-phospho-p53 (pSer15) antibody produced in rabbit, affinity isolated antibody
SAB4503953
Anti-phospho-p53 (pSer20) antibody produced in rabbit, affinity isolated antibody
SAB4504500
Anti-phospho-p53 (pSer315) antibody produced in rabbit, affinity isolated antibody
SAB4504501
Anti-phospho-p53 (pSer33) antibody produced in rabbit, affinity isolated antibody
SAB4504592
Anti-phospho-p53 (pSer366) antibody produced in rabbit, affinity isolated antibody
SAB4504502
Anti-phospho-p53 (pSer37) antibody produced in rabbit, affinity isolated antibody
SAB4504593
Anti-phospho-p53 (pSer378) antibody produced in rabbit, affinity isolated antibody
P8982 Anti-phospho-p53 (pSer392) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503954
Anti-phospho-p53 (pSer392) antibody produced in rabbit, affinity isolated antibody
SAB4504503
Anti-phospho-p53 (pSer46) antibody produced in rabbit, affinity isolated antibody
SAB4504504
Anti-phospho-p53 (pSer6) antibody produced in rabbit, affinity isolated antibody
SAB4504505
Anti-phospho-p53 (pSer9) antibody produced in rabbit, affinity isolated antibody
SAB4504506
Anti-phospho-p53 (pThr18) antibody produced in rabbit, affinity isolated antibody
SAB4504591
Anti-phospho-p53 (pThr387) antibody produced in rabbit, affinity isolated antibody
SAB1306688 ANTI-PHOSPHO-P53(S20) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305629 ANTI-PHOSPHO-P53(S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306606
ANTI-PHOSPHO-P53(S9) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306609 ANTI-PHOSPHO-P53(T18) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305574 ANTI-PHOSPHO-P53(T55) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300079
Anti-phospho-TP53 (pSer315) antibody produced in rabbit, affinity isolated antibody
SAB4300076
Anti-phospho-TP53 (pSer33) antibody produced in rabbit, affinity isolated antibody
SAB4300077
Anti-phospho-TP53 (pSer37) antibody produced in rabbit, affinity isolated antibody
SAB4300078
Anti-phospho-TP53 (pSer46) antibody produced in rabbit, affinity isolated antibody
SAB4300072
Anti-phospho-TP53 (pSer6) antibody produced in rabbit, affinity isolated antibody
SAB4300074
Anti-phospho-TP53 (pThr18) antibody produced in rabbit, affinity isolated antibody
D7696
Demethoxycurcumin, ≥98% (HPLC)
C20H18O5
M3068 Monoclonal Anti-MGMT antibody produced in mouse, clone MGMT-214, purified immunoglobulin, buffered aqueous solution