EMAIL THIS PAGE TO A FRIEND
Molecular oral microbiology
Streptococcus gordonii DL1 adhesin SspB V-region mediates coaggregation via receptor polysaccharide of Actinomyces oris T14V.
PMID 25965671
Abstract
Streptococcus gordonii SspA and SspB proteins, members of the antigen I/II (AgI/II) family of Streptococcus adhesins, mediate adherence to cysteine-rich scavenger glycoprotein gp340 and cells of other oral microbial species. In this article we investigated further the mechanism of coaggregation between S. gordonii DL1 and Actinomyces oris T14V. Previous mutational analysis of S. gordonii suggested that SspB was necessary for coaggregation with A. oris T14V. We have confirmed this by showing that Lactococcus lactis surrogate host cells expressing SspB coaggregated with A. oris T14V and PK606 cells, while L. lactis cells expressing SspA did not. Coaggregation occurred independently of expression of A. oris type 1 (FimP) or type 2 (FimA) fimbriae. Polysaccharide was prepared from cells of A. oris T14V and found to contain 1,4-, 4,6- and 3,4-linked glucose, 1,4-linked mannose, and 2,4-linked galactose residues. When immobilized onto plastic wells this polysaccharide supported binding of L. lactis expressing SspB, but not binding of L. lactis expressing other AgI/II family proteins. Purified recombinant NAVP region of SspB, comprising amino acid (aa) residues 41-847, bound A. oris polysaccharide but the C-domain (932-1470 aa residues) did not. A site-directed deletion of 29 aa residues (Δ691-718) close to the predicted binding cleft within the SspB V-region ablated binding of the NAVP region to polysaccharide. These results infer that the V-region head of SspB recognizes an actinomyces polysaccharide ligand, so further characterizing a lectin-like coaggregation mechanism occurring between two important primary colonizers.