EMAIL THIS PAGE TO A FRIEND
Oncotarget
Stromal androgen receptor regulates the composition of the microenvironment to influence prostate cancer outcome.
PMID 25965833
Abstract
Androgen receptor (AR) signaling in stromal cells is important in prostate cancer, yet the mechanisms underpinning stromal AR contribution to disease development and progression remain unclear. Using patient-matched benign and malignant prostate samples, we show a significant association between low AR levels in cancer associated stroma and increased prostate cancer-related death at one, three and five years post-diganosis, and in tissue recombination models with primary prostate cancer cells that low stromal AR decreases castration-induced apoptosis. AR-regulation was found to be different in primary human fibroblasts isolated from adjacent to cancerous and non-cancerous prostate epithelia, and to represent altered activation of myofibroblast pathways involved in cell cycle, adhesion, migration, and the extracellular matrix (ECM). Without AR signaling, the fibroblast-derived ECM loses the capacity to promote attachment of both myofibroblasts and cancer cells, is less able to prevent cell-matrix disruption, and is less likely to impede cancer cell invasion. AR signaling in prostate cancer stroma appears therefore to alter patient outcome by maintaining an ECM microenvironment inhibitory to cancer cell invasion. This paper provides comprehensive insight into AR signaling in the non-epithelial prostate microenvironment, and a resource from which the prognostic and therapeutic implications of stromal AR levels can be further explored.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
660124
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, 97%
C36H24N2
699195
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, sublimed grade, 99.9% trace metals basis
C36H24N2
40205
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade PURANAL (Honeywell 17814)
H5NO
40291
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade VLSI PURANAL (Honeywell 17605)
H5NO
40321
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade SLSI PURANAL (Honeywell 17301)
H5NO
01-4362
Ammonium hydroxide solution, 28% NH3 in H2O, suitable for determination of toxic metals
H5NO
SAB1409018
Anti-AKR1B1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV48180
Anti-AKR1B1 antibody produced in rabbit, IgG fraction of antiserum
SAB2100091
Anti-AKR1B1 antibody produced in rabbit, affinity isolated antibody
HPA026425
Anti-AKR1B1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410404
Anti-AKR1B1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1410405
Anti-AKR1B1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4500944
Anti-AKR1B1, C-Terminal antibody produced in rabbit, affinity isolated antibody
GW22855 Anti-Amphiregulin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4300382
Anti-AR (Ab-650) antibody produced in rabbit, affinity isolated antibody
SAB2100139
Anti-AR antibody produced in rabbit, affinity isolated antibody
SAB1405478
Anti-AREG antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA008720
Anti-AREG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410727
Anti-AREG antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1400100
Anti-FDXR antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1400101
Anti-FDXR antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2100807
Anti-FDXR antibody produced in rabbit, affinity isolated antibody
SAB1405814
Anti-FKBP5 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100820
Anti-FKBP5 antibody produced in rabbit, affinity isolated antibody
HPA031092
Anti-FKBP5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA031093
Anti-FKBP5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA031095
Anti-FKBP5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401521
Anti-TUBA1B antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1402112
Monoclonal Anti-AKR1B1 antibody produced in mouse, clone 2D12, purified immunoglobulin, buffered aqueous solution
WH0000367M1
Monoclonal Anti-AR antibody produced in mouse, clone 1G3, purified immunoglobulin, buffered aqueous solution