EMAIL THIS PAGE TO A FRIEND
Lipids
Fish Oil and Olive Oil Supplementation in Late Pregnancy and Lactation Differentially Affect Oxidative Stress and Inflammation in Sows and Piglets.
PMID 25968335
Abstract
This study was conducted to compare the effects of fish oil and olive oil supplementation in late pregnancy and during lactation on oxidative stress and inflammation in sows and their piglets. A total of 24 sows were fed a basal diet supplemented with additional corn starch (CON), fish oil (FO) or olive oil (OO). Sows fed an OO diet during late gestation had a higher piglet birth weight compared with CON-fed and FO-fed sows (Pxa0