EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
Synergistic activity of combination therapy with PEGylated pemetrexed and gemcitabine for an effective cancer treatment.
PMID 25968494
Abstract
Combination therapy in cancer is now opted as a potential therapeutic strategy for cancer treatment. However, effective delivery of drugs in combination at the tumor site is marred by low bioavailability and systemic toxicity of individual drugs. Polymer therapeutics is indeed an upcoming approach for the combinational drug delivery in favor of better cancer management. Hence, the objective of our investigation was to develop a dual drug PEGylated system that carries two chemotherapeutic drugs simultaneously for effective treatment of cancer. In this regard, we have synthesized Pem-PEG-Gem, wherein pemetrexed (Pem) and gemcitabine (Gem) are conjugated to a heterobifunctional polyethylene glycol (PEG) polymer for the effective treatment of Non-Small Cell Lung Cancer (NSCLC). Our results demonstrate enhanced bioavailability of the individual drugs in Pem-PEG-Gem in comparison with the drugs in their native form. The developed Pem-PEG-Gem showed enhanced cell death with respect to their native counterparts when treated singly or in combination against NSCLC cells. This might be attributed to better cellular internalization through the process of macropinocytosis and synergistic cytotoxic action of Pem-PEG-Gem in NSCLC cells. Hence, we propose the above dual drug based polymer therapeutic approach suitable for better clinical application in the treatment of NSCLC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G6423
Gemcitabine hydrochloride, ≥98% (HPLC)
C9H11F2N3O4 · HCl