EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Efficient Semitransparent Perovskite Solar Cells with Graphene Electrodes.
PMID 25969400
Abstract
Semitransparent perovskite solar cells are prepared by laminating graphene transparent electrodes on the top for the first time. The device performance is optimized by improving the conductivity of the graphene electrodes and the contact between the graphene and the perovskite active layers during the lamination process. The devices show high power conversion efficiencies when they are illuminated from both sides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
900442
Graphene/PEDOT:PSS hybrid ink, dispersion in DMF