EMAIL THIS PAGE TO A FRIEND
Oncotarget
The interplay between TEAD4 and KLF5 promotes breast cancer partially through inhibiting the transcription of p27Kip1.
PMID 25970772
Abstract
Growing evidence suggests that YAP/TAZ are mediators of the Hippo pathway and promote breast cancer. However, the roles of YAP/TAZ transcription factor partners TEADs in breast cancer remain unclear. Here we found that TEAD4 was expressed in breast cancer cell lines, especially in triple negative breast cancers (TNBC) cell lines. TEAD4 binds to KLF5. Knockdown of either TEAD4 or KLF5 in HCC1937 and HCC1806 cells induced the expression of CDK inhibitor p27. Depletion of either TEAD4 or KLF5 activated the p27 gene promoter and increased the p27 mRNA levels. Depletion of p27 partially prevents growth inhibition caused by TEAD4 and KLF5 knockdown. TEAD4 overexpression stimulated proliferation in vitro and tumor growth in mice, while stable knockdown of TEAD4 inhibited proliferation in vitro and tumor growth in mice. Thus TEAD4 and KLF5, in collaboration, promoted TNBC cell proliferation and tumor growth in part by inhibiting p27 gene transcription. TEAD4 is a potential target and biomarker for the development of novel therapeutics for breast cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7781
Anti-Glutathione-S-Transferase (GST) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA040398
Anti-KLF5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701988
Anti-KLF5 antibody produced in rabbit
AV39521
Anti-TEAD1 (AB1) antibody produced in rabbit, affinity isolated antibody
SAB2102401
Anti-TEAD1 antibody produced in rabbit, affinity isolated antibody
SAB2501231
Anti-TEAD2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2102402
Anti-TEAD2 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2104126
Anti-TEAD2 antibody produced in rabbit, affinity isolated antibody
SAB4503373
Anti-TEAD2 antibody produced in rabbit, affinity isolated antibody
AV33429
Anti-TEAD3 (AB1) antibody produced in rabbit, affinity isolated antibody
AV38278
Anti-TEAD3 (AB2) antibody produced in rabbit, affinity isolated antibody
AV38277
Anti-TEAD3 antibody produced in rabbit, affinity isolated antibody
HPA028906
Anti-TEAD3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102987
Anti-TEAD3 antibody produced in rabbit, affinity isolated antibody
SAB1406521
Anti-TEAD4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV38276
Anti-TEAD4 antibody produced in rabbit, affinity isolated antibody
SAB2102403
Anti-TEAD4 antibody produced in rabbit, affinity isolated antibody
HPA056896
Anti-TEAD4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106301
Anti-TEAD4 antibody produced in rabbit, affinity isolated antibody
SAB1411339
Anti-TEAD4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
F9291 Monoclonal ANTI-FLAG® BioM2 antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
SAB4200338
Monoclonal Anti-KLF5 antibody produced in mouse, ~1.0 mg/mL, clone KLF5-55, purified immunoglobulin
SAB1402132
Monoclonal Anti-KLF5, (C-terminal) antibody produced in mouse, clone 2G12, purified immunoglobulin, buffered aqueous solution
WH0008463M1 Monoclonal Anti-TEAD2 antibody produced in mouse, clone 2D5, purified immunoglobulin, buffered aqueous solution
WH0007005M1 Monoclonal Anti-TEAD3 antibody produced in mouse, clone 1C4, purified immunoglobulin, buffered aqueous solution
WH0007004M1
Monoclonal Anti-TEAD4 antibody produced in mouse, clone 5H3, purified immunoglobulin, buffered aqueous solution
SAB1404457
Monoclonal Anti-TEAD4, (N-terminal) antibody produced in mouse, clone 1D10, purified immunoglobulin, buffered aqueous solution