EMAIL THIS PAGE TO A FRIEND
Oncotarget
Ccdc6 knock-in mice develop thyroid hyperplasia associated to an enhanced CREB1 activity.
PMID 25970781
Abstract
CCDC6 was originally identified upon rearrangement with RET in human thyroid papillary carcinomas generating the RET/PTC1 oncogene. We have previously reported that CCDC6 interacts with CREB1 and represses its transcriptional activity. Since the function of at least one allele of CCDC6 is lost following RET/PTC1 rearrangements, we aimed at the generation of mice, carrying a CCDC6 mutant gene. Previous studies suggested that the coiled-coil domain of CCDC6, mainly encoded by human exon 2, is required for the protein function. Therefore, we engineered a murine Ccdc6 construct, carrying a deletion of the exon 2, that was able to exert only a mild repression on CREB1 transcriptional activity, with respect to the wild type Ccdc6. Subsequently, we generated Ccdc6-ex2 knock-in mice. These mice developed thyroid hyperplasia associated with an enhanced CREB1 activity and an increased expression of the CREB-1 regulated genes. These results strongly support a CCDC6 promoting role, ascribed to its functional impairment, in the development of thyroid papillary carcinomas harboring the RET/PTC1 oncogene.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1409834
Anti-CCDC6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA019049
Anti-CCDC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA019051
Anti-CCDC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2107667
Anti-CCDC6 antibody produced in rabbit, affinity isolated antibody
SAB1411455
Anti-CCDC6 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4500441
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4500442
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4500443
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4500444
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4500445
Anti-CREB antibody produced in rabbit, affinity isolated antibody
SAB4300519
Anti-CREB1 (Ab-129) antibody produced in rabbit, affinity isolated antibody
SAB4300333
Anti-CREB1 (Ab-133) antibody produced in rabbit, affinity isolated antibody
SAB4300546
Anti-CREB1 (Ab-142) antibody produced in rabbit, affinity isolated antibody
AV37464
Anti-CREB1 (AB1) antibody produced in rabbit, affinity isolated antibody
AV01026
Anti-CREB1 (AB2) antibody produced in rabbit, affinity isolated antibody
AV31264
Anti-CREB1 antibody produced in rabbit, IgG fraction of antiserum
HPA019150
Anti-CREB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103540
Anti-CREB1 antibody produced in rabbit, affinity isolated antibody
SAB4503908
Anti-phospho-CREB (pSer121) antibody produced in rabbit, affinity isolated antibody
SAB4504375
Anti-phospho-CREB (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB4503757
Anti-phospho-CREB (pSer142) antibody produced in rabbit, affinity isolated antibody
SAB4504577
Anti-phospho-CREB (pThr100) antibody produced in rabbit, affinity isolated antibody
SAB1306301 ANTI-PHOSPHO-CREB(S133) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300217
Anti-phospho-CREB1 (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4300040
Anti-phospho-CREB1 (pSer133) antibody produced in rabbit, affinity isolated antibody
H0412 Anti-phospho-Histone H3 (pSer10) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4503877
Anti-phospho-Histone H3 (pSer28) antibody produced in rabbit, affinity isolated antibody
SAB4503876
Anti-phospho-Histone H3 (pThr3) antibody produced in rabbit, affinity isolated antibody
WH0008030M3
Monoclonal Anti-CCDC6 antibody produced in mouse, clone 5D11, purified immunoglobulin, buffered aqueous solution
WH0001385M1
Monoclonal Anti-CREB1 antibody produced in mouse, clone 3F1-1B2, purified immunoglobulin, buffered aqueous solution