EMAIL THIS PAGE TO A FRIEND
Journal of fluorescence
TEMPO-Attached Pre-fluorescent Probes Based on Pyridinium Fluorophores.
PMID 25972068
Abstract
The photophysical behavior of three pyridinium-derived fluorophores, the N-aryl-2,4,6-triphenylpyridinium, the N-aryl-5,6-dihydro-2,4-diphenylbenzo[h]quinolinium and the N-aryl-5,6,8,9-tetrahydro-7-phenyldibenzo[c,h]acridinium perchlorates, was investigated. Comparison of their fluorescence quantum yields led to the preparation of a novel, more sensitive pyridinium-based, TEMPO-attached prefluorescent probe for H-abstraction processes, the N-{4-[4-(N-oxyl-2,2,6,6-tetramethylpiperidinyl)carbonylamino]phenyl}-5,6,8,9-tetrahydro-7-phenyldibenzo[c,h]-acridinium perchlorate.