EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
The N-ethylmaleimide-sensitive factor and dysbindin interact to modulate synaptic plasticity.
PMID 25972187
Abstract
Dysbindin is a schizophrenia susceptibility factor and subunit of the biogenesis of lysosome-related organelles complex 1 (BLOC-1) required for lysosome-related organelle biogenesis, and in neurons, synaptic vesicle assembly, neurotransmission, and plasticity. Protein networks, or interactomes, downstream of dysbindin/BLOC-1 remain partially explored despite their potential to illuminate neurodevelopmental disorder mechanisms. Here, we conducted a proteome-wide search for polypeptides whose cellular content is sensitive to dysbindin/BLOC-1 loss of function. We identified components of the vesicle fusion machinery as factors downregulated in dysbindin/BLOC-1 deficiency in neuroectodermal cells and iPSC-derived human neurons, among them the N-ethylmaleimide-sensitive factor (NSF). Human dysbindin/BLOC-1 coprecipitates with NSF and vice versa, and both proteins colocalized in a Drosophila model synapse. To test the hypothesis that NSF and dysbindin/BLOC-1 participate in a pathway-regulating synaptic function, we examined the role for NSF in dysbindin/BLOC-1-dependent synaptic homeostatic plasticity in Drosophila. As previously described, we found that mutations in dysbindin precluded homeostatic synaptic plasticity elicited by acute blockage of postsynaptic receptors. This dysbindin mutant phenotype is fully rescued by presynaptic expression of either dysbindin or Drosophila NSF. However, neither reduction of NSF alone or in combination with dysbindin haploinsufficiency impaired homeostatic synaptic plasticity. Our results demonstrate that dysbindin/BLOC-1 expression defects result in altered cellular content of proteins of the vesicle fusion apparatus and therefore influence synaptic plasticity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F4799 3X FLAG® Peptide, lyophilized powder
C120H169N31O49S
HPA039928
Anti-BLOC1S6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1300674
Anti-DTNBP1 (C-term) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1300673
Anti-DTNBP1 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1401882
Anti-DTNBP1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA028053
Anti-DTNBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA029615
Anti-DTNBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA029616
Anti-DTNBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3
SAB4200486
Anti-Dysbindin (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4200409
Anti-Dysbindin antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB2500742
Anti-Pallidin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1400488
Anti-PLDN antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2101826
Anti-PLDN antibody produced in rabbit, affinity isolated antibody
SAB2105000
Anti-PLDN, (N-terminal) antibody produced in rabbit, affinity isolated antibody
P0078
Anti-PMP22 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4502217
Anti-PMP22, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406663
Anti-STX7 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001467
Anti-STX7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401426
Anti-STX7 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2107605
Anti-STX7 antibody produced in rabbit, affinity isolated antibody
SAB2700571
Anti-STXBP5 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
S4819
Anti-Syntaxin 7 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2501035
Anti-Thioredoxin Reductase 1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1400287
Anti-TXNRD1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA001395
Anti-TXNRD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411372
Anti-TXNRD1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
186260
Ethanesulfonic acid, 95%
C2H6O3S
471550
Ethanesulfonic acid solution, 70 wt. % in H2O
C2H6O3S
SAB1401883
Monoclonal Anti-DTNBP1 antibody produced in mouse, clone 1B4, purified immunoglobulin, buffered aqueous solution
SAB1405011
Monoclonal Anti-PLDN antibody produced in mouse, clone 3D2, purified immunoglobulin, buffered aqueous solution