EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Poly(inosinic-cytidylic) acid-triggered exacerbation of experimental asthma depends on IL-17A produced by NK cells.
PMID 25972482
Abstract
Viral infection of the respiratory tract represents the major cause of acute asthma exacerbations. dsRNA is produced as an intermediate during replication of respiratory viruses and triggers immune responses via TLR3. This study aimed at clarifying the mechanisms underlying TLR3 triggered exacerbation of experimental allergic asthma. The TLR3 ligand poly(inosinic-cytidylic) acid was applied intranasally to mice with already established experimental allergic asthma. Airway inflammation, cytokine expression, mucus production, and airway reactivity was assessed in wild-type, IL-17A, or IL-23p19-deficient, and in NK cell-depleted mice. Local application of poly(inosinic-cytidylic) acid exacerbated experimental allergic asthma in mice as characterized by enhanced release of proinflammatory cytokines, aggravated airway inflammation, and increased mucus production together with pronounced airway hyperresponsiveness. This was further associated with augmented production of IL-17 by Th17 cells and NK cells. Whereas experimental exacerbation could be induced in IL-23p19-deficient mice lacking mature, proinflammatory Th17 cells, this was not possible in mice lacking IL-17A or in NK cell-depleted animals. These experiments indicate a central role for IL-17 derived from NK cells but not from Th17 cells in the pathogenesis of virus-triggered exacerbation of experimental asthma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1918
Pancuronium bromide
C35H60Br2N2O4