EMAIL THIS PAGE TO A FRIEND
Cell death & disease
RGD-modifided oncolytic adenovirus exhibited potent cytotoxic effect on CAR-negative bladder cancer-initiating cells.
PMID 25973680
Abstract
Cancer-initiating cell (CIC) is critical in cancer development, maintenance and recurrence. The reverse expression pattern of coxsackie and adenovirus receptor (CAR) and αν integrin in bladder cancer decreases the infection efficiency of adenovirus. We constructed Arg-Gly-Asp (RGD)-modified oncolytic adenovirus, carrying EGFP or TNF-related apoptosis-inducing ligand (TRAIL) gene (Onco(Ad).RGD-hTERT-EGFP/TRAIL), and applied them to CAR-negative bladder cancer T24 cells and cancer-initiating T24 sphere cells. Onco(Ad).RGD-hTERT-EGFP had enhanced infection ability and cytotoxic effect on T24 cells and T24 sphere cells, but little cytoxicity on normal urothelial SV-HUC-1 cells compared with the unmodified virus Onco(Ad).hTERT-EGFP. Notably, Onco(Ad).RGD-hTERT-TRAIL induced apoptosis in T24 cells and T24 sphere cells. Furthermore, it completely inhibited xenograft initiation established by the oncolytic adenovirus-pretreated T24 sphere cells, and significantly suppressed tumor growth by intratumoral injection. These results provided a promising therapeutic strategy for CAR-negative bladder cancer through targeting CICs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV100591
Anti-NANOG (AB1) antibody produced in rabbit, affinity isolated antibody
AV47988
Anti-NANOG (AB2) antibody produced in rabbit, affinity isolated antibody
SAB2500670
Anti-NANOG antibody produced in goat, affinity isolated antibody, buffered aqueous solution
N4413 Anti-Nanog antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4300680
Anti-Nanog antibody produced in rabbit, affinity isolated antibody
SAB3500389
Anti-NANOG antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104002
Anti-NANOG antibody produced in rabbit, affinity isolated antibody
SAB2104004
Anti-NANOG, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2500671
Anti-NANOG, C-Terminal antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1411780
ANTI-RCV1 antibody produced in mouse, clone 5A6, purified immunoglobulin, buffered aqueous solution
SAB1411781
ANTI-RCV1 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution
SAB1411794
ANTI-RCV1 antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
SAB1411795
ANTI-RCV1 antibody produced in mouse, clone 4C10, purified immunoglobulin, buffered aqueous solution
SAB1411804
ANTI-RCV1 antibody produced in mouse, clone 4D9, purified immunoglobulin, buffered aqueous solution
SAB1411805
ANTI-RCV1 antibody produced in mouse, clone 2B3, purified immunoglobulin, buffered aqueous solution
SAB1411862
ANTI-RCV1 antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1411863
ANTI-RCV1 antibody produced in mouse, clone 4E2, purified immunoglobulin, buffered aqueous solution
SAB1411864
ANTI-RCV1 antibody produced in mouse, clone 4A8, purified immunoglobulin, buffered aqueous solution
SAB1411865
ANTI-RCV1 antibody produced in mouse, clone 2B2, purified immunoglobulin, buffered aqueous solution
SAB1411878
ANTI-RCV1 antibody produced in mouse, clone 1B9, purified immunoglobulin, buffered aqueous solution
SAB1411879
ANTI-RCV1 antibody produced in mouse, clone 1B7, purified immunoglobulin, buffered aqueous solution
AV38196
Anti-RCV1 antibody produced in rabbit, affinity isolated antibody
SAB1406380
Anti-RCVRN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101971
Anti-RCVRN antibody produced in rabbit, affinity isolated antibody
SAB4200309
Anti-Recoverin (C-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
SAB4502604
Anti-Recoverin antibody produced in rabbit, affinity isolated antibody
N3038
Monoclonal Anti-Nanog antibody produced in mouse, ~2 mg/mL, clone NNG-811, purified immunoglobulin, buffered aqueous solution
WH0079923M1
Monoclonal Anti-NANOG antibody produced in mouse, clone 2C11, purified immunoglobulin, buffered aqueous solution
SAB1405280
Monoclonal Anti-NANOG antibody produced in mouse, clone 2E11, purified immunoglobulin, buffered aqueous solution
WH0005957M1
Monoclonal Anti-RCV1 antibody produced in mouse, clone 4C6, purified immunoglobulin, buffered aqueous solution