EMAIL THIS PAGE TO A FRIEND
Journal of refractive surgery (Thorofare, N.J. : 1995)
Induced inflammation and apoptosis in femtosecond laser-assisted capsulotomies and manual capsulorhexes: an immunohistochemical study.
PMID 25974966
Abstract
To evaluate cellular inflammation and apoptosis induced in the central portion of capsulorhexes/capsulotomies during cataract surgery, comparing a conventional manual technique and a femtosecond laser-assisted procedure at different energy settings using two laser systems. Fifty-six capsulorhexes/capsulotomies were divided into four groups: the manual group (14 capsulorhexes) performed with the manual technique; the 7.0-µJ group (14 capsulotomies) (LensAR laser system; Lensar, Inc., Orlando, FL); the 10-µJ group (14 capsulotomies) (LenSx laser system; Alcon Laboratories, Inc., Fort Worth, TX); and the 13.0-µJ group (14 capsulotomies) (LenSx laser system). All samples were stained for cellular apoptosis analysis (TUNEL assay) and cellular induced inflammation (NF-κB). One-way analysis of variance indicated a statistically significant difference in the percentage of NF-κB and TUNEL positive cells between the four groups, (F [3.52] = 14.717, P < .001) and (F [3.52] = 139.561, P < .001), respectively. Post-hoc analysis indicated a statistically significant difference in the percentage of NF-κB positive cells between the 13.0-µJ group and the manual, 7.0-µJ, and 10-µJ groups (P < .001, = .037, and < .001, respectively). Post-hoc analysis of differences in TUNEL positive cells indicated a significant difference between the 7.0-µJ and 10-µJ groups (P <.017) and between the 13.0-µJ group and the manual, 7.0-µJ, and 10-µJ groups (P < .001, < .001, and < .001, respectively). The results show a higher percentage of NF-κB and TUNEL positive cells in the 13.0-µJ group compared to the 7.0-µJ, 10-µJ, and manual groups. Therefore, inflammatory response and cell death increased at increasing energies. An effective capsulotomy in femtosecond laser-assisted cataract surgery with minimal detrimental apoptotic and inflammatory effects is possible if the laser system is set to use the minimum energy level.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501989
Anti-NF-κB p105 antibody produced in rabbit, affinity isolated antibody
SAB4300301
Anti-NFKB1 (Ab-337) antibody produced in rabbit, affinity isolated antibody
SAB4300302
Anti-NFKB1 (Ab-893) antibody produced in rabbit, affinity isolated antibody
SAB4300303
Anti-NFKB1 (Ab-907) antibody produced in rabbit, affinity isolated antibody
SAB4300554
Anti-NFKB1 (Ab-927) antibody produced in rabbit, affinity isolated antibody
SAB4300498
Anti-NFKB1 (Ab-932) antibody produced in rabbit, affinity isolated antibody
GW21167 Anti-NFKB1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1406182
Anti-NFKB1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV38413
Anti-NFKB1 antibody produced in rabbit, affinity isolated antibody
HPA027305
Anti-NFKB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103906
Anti-NFKB1 antibody produced in rabbit, affinity isolated antibody
N8523
Monoclonal Anti-NF-κB antibody produced in mouse, clone NF-12, ascites fluid
WH0004790M1
Monoclonal Anti-NFKB1 antibody produced in mouse, clone 2E6, purified immunoglobulin, buffered aqueous solution
SAB1404125
Monoclonal Anti-NFKB1, (C-terminal) antibody produced in mouse, clone 4A11, purified immunoglobulin, buffered aqueous solution
PLA0214
Rabbit anti-NF-kappaB1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.