EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Cryptic noninnocence: FeNO corroles in a new light.
PMID 25978788
Abstract
Multiple lines of evidence, including electronic absorption spectroscopy, infrared spectroscopy, and broken-symmetry DFT calculations, indicate that the well-known FeNO corroles, long assumed to be {FeNO}(6) complexes, are in fact better described as {FeNO}(7)-(corrole˙(2-)).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W338605
Pyrrole, ≥98%, FCC, FG
C4H5N