EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Trans-regulation of Syndecan Functions by Hetero-oligomerization.
PMID 25979339
Abstract
Syndecans, a family of transmembrane heparansulfate proteoglycans, are known to interact through their transmembrane domains to form non-covalently linked homodimers, a process essential for their individual functions. Because all syndecan transmembrane domains are highly conserved and thus might mediate interactions between different members of the syndecan family, we investigated syndecan interactions in detail. All recombinant syndecan-2 and -4 protein variants containing the transmembrane domain formed not only sodium dodecyl sulfate (SDS)-resistant homodimers but also SDS-resistant heterodimers. Biochemical and structural data revealed that recombinant syndecan-2 and -4 formed intermolecular interactions in vitro, and the GXXXG motif in transmembrane domain mediated this interaction. When exogenously expressed in rat embryonic fibroblasts, syndecan-2 interacted with syndecan-4 and vice versa. Furthermore, bimolecular fluorescence complementation-based assay demonstrated specific hetero-molecular interactions between syndecan-2 and -4, supporting hetero-oligomer formation of syndecans in vivo. Interestingly, hetero-oligomerization significantly reduced syndecan-4-mediated cellular processes such as protein kinase Cα activation and protein kinase Cα-mediated cell adhesion as well as syndecan-2-mediated tumorigenic activities in colon cancer cells such as migration and anchorage-independent growth. Taken together, these data provide evidence that hetero-oligomerization produces distinct syndecan functions and offer insights into the underlying signaling mechanisms of syndecans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4502553
Anti-Paxillin antibody produced in rabbit, affinity isolated antibody
SAB4300384
Anti-PXN (Ab-118) antibody produced in rabbit, affinity isolated antibody
SAB4300459
Anti-PXN (Ab-31) antibody produced in rabbit, affinity isolated antibody
SAB4300592
Anti-PXN (Ab-88) antibody produced in rabbit, affinity isolated antibody
SAB1406355
Anti-PXN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401357
Anti-SDC2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1409636
Anti-SDC2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1406419
Anti-SDC4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA005716
Anti-SDC4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408631
Anti-SDC4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1305542
MONOCLONAL ANTI-CD138 (C-TERMINAL) antibody produced in mouse, clone 587CT7.3.6.5, crude ascites, buffered aqueous solution
P1093
Monoclonal Anti-Paxillin antibody produced in mouse, clone PXC-10, ascites fluid
SAB1401358
Monoclonal Anti-SDC2 antibody produced in mouse, clone 1B2, purified immunoglobulin, buffered aqueous solution