EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Unique Features of Human Protein Arginine Methyltransferase 9 (PRMT9) and Its Substrate RNA Splicing Factor SF3B2.
PMID 25979344
Abstract
Human protein arginine methyltransferase (PRMT) 9 symmetrically dimethylates arginine residues on splicing factor SF3B2 (SAP145) and has been functionally linked to the regulation of alternative splicing of pre-mRNA. Site-directed mutagenesis studies on this enzyme and its substrate had revealed essential unique residues in the double E loop and the importance of the C-terminal duplicated methyltransferase domain. In contrast to what had been observed with other PRMTs and their physiological substrates, a peptide containing the methylatable Arg-508 of SF3B2 was not recognized by PRMT9 in vitro. Although amino acid substitutions of residues surrounding Arg-508 had no great effect on PRMT9 recognition of SF3B2, moving the arginine residue within this sequence abolished methylation. PRMT9 and PRMT5 are the only known mammalian enzymes capable of forming symmetric dimethylarginine (SDMA) residues as type II PRMTs. We demonstrate here that the specificity of these enzymes for their substrates is distinct and not redundant. The loss of PRMT5 activity in mouse embryo fibroblasts results in almost complete loss of SDMA, suggesting that PRMT5 is the primary SDMA-forming enzyme in these cells. PRMT9, with its duplicated methyltransferase domain and conserved sequence in the double E loop, appears to have a unique structure and specificity among PRMTs for methylating SF3B2 and potentially other polypeptides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9393
Ampicillin, anhydrous, 96.0-100.5% (anhydrous basis)
C16H19N3O4S
A1593
Ampicillin, meets USP testing specifications
C16H19N3O4S · 3H2O
B2261
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
C27H28N6O · 3HCl · xH2O
14533
bisBenzimide H 33342 trihydrochloride, for fluorescence, ≥97.0% (HPLC)
C27H28N6O · 3HCl · xH2O
11020
DL-Arginine, ≥97.0% (NT)
C6H14N4O2
H7755
Histone from calf thymus, Type II-AS
M1891 Myelin Basic Protein bovine, ≥90% (SDS-PAGE), lyophilized powder
D0390
NG,NG′-Dimethyl-L-arginine di(p-hydroxyazobenzene-p′-sulfonate) salt, ≥99% (TLC)
C8H18N4O2 · 2C12H10N2O4S
D4268
NG,NG-Dimethylarginine dihydrochloride
C8H18N4O2 · 2HCl
735914
Octylphosphonic acid, 97%
C8H19O3P
P0657
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder
C8H6O2
P1378
Phthaldialdehyde, ≥97% (HPLC), powder (may contain lumps)
C8H6O2
79760
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
C8H6O2
RDD004
PIPES, anhydrous, free-flowing, Redi-Dri, ≥99%
C8H18N2O6S2
P6757
PIPES, ≥99% (titration)
C8H18N2O6S2
RES0703P-A7
PIPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O6S2
P1851
PIPES, BioPerformance Certified, suitable for cell culture, ≥99%
C8H18N2O6S2
P8203
PIPES, BioXtra, ≥99% (titration)
C8H18N2O6S2
80635
PIPES, BioXtra, for molecular biology, ≥99.5% (T)
C8H18N2O6S2
244856
Potassium, ingot, 99.95% trace metals basis
K
244864
Potassium, chunks (in mineral oil), 98% trace metals basis
K
679909
Potassium, cubes (in mineral oil), L × W × H 40 mm × 30 mm × 20 mm, 99.5% trace metals basis
K
215813
Potassium hydride, 30 wt % dispersion in mineral oil
HK
708860
Potassium hydride, in paraffin
HK