EMAIL THIS PAGE TO A FRIEND
Annales pharmaceutiques francaises
Forced degradation studies, and effect of surfactants and titanium dioxide on the photostability of paliperidone by HPLC.
PMID 25980637
Abstract
Forced degradation study of paliperidone under hydrolytic, oxidative, thermal and photolytic stress conditions was conducted using HPLC. The drug was found to be labile under hydrolytic, oxidative and photolytic stress conditions; whereas, it was stable under dry heat stress conditions. Effect of anionic, cationic and non-ionic surfactants applied to the concentration exceeding critical micellar concentration on the photostability of paliperidone was also studied by exposing the samples to sunlight for 72h. Major degradation of the drug was found in presence of cationic and non-ionic surfactants. Effect of titanium dioxide on the photo-degradation of paliperidone in solution state was also studied and it was found that 53% of the drug was degraded after 72h of exposure to sunlight. A common degradation peak was observed in oxidative and TiO2 photocatalysed samples. This peak may be due to the generation of N-oxide of paliperidone. The same degradation peak was also observed in all other photostability samples. Chromatographic separation of drug and its degradation products was achieved on an Alltima C8 (250mm×4.6mm, 5μm) analytical column, using a mobile phase consisting of acetonitrile-ammonium acetate buffer with 0.2% triethylamine (pH 3.5; 20mM) (60:40, v/v) at a flow rate of 1mL/min. Quantification was performed with UV detection at 280nm. The method was validated as per ICH guidelines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ARK2188
Hexadecyltrimethylammonium bromide, PharmaGrade, USP/NF, Manufactured under appropriate controls for pharma or biopharmaceutical production.
C19H42BrN
H9151
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
C19H42BrN
52365
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
C19H42BrN
H6269
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
C19H42BrN
H5882
Hexadecyltrimethylammonium bromide, ≥98%
C19H42BrN
P5811
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
H3O4P
466123
Phosphoric acid, crystalline, ≥99.999% trace metals basis
H3O4P
452289
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
H3O4P
345245
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
H3O4P
79617
Phosphoric acid, BioUltra, ≥85% (T)
H3O4P
40278
Phosphoric acid, semiconductor grade MOS PURANAL (Honeywell 17938), ≥85%
H3O4P
40266
Phosphoric acid, semiconductor grade PURANAL (Honeywell 17861), ≥85%
H3O4P
612162
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
H3O4P
699608
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
H3O4P
698822
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
H3O4P
W290017
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
H3O4P
609781
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
H316O4P
T0886
Triethylamine, ≥99%
C6H15N
90335
Triethylamine, BioUltra, ≥99.5% (GC)
C6H15N
90337
Triethylamine, for amino acid analysis, ≥99.5% (GC)
C6H15N
90338
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
C6H15N
90340
Triethylamine, puriss. p.a., ≥99.5% (GC)
C6H15N
16304
Triethylamine, purum, ≥99% (GC)
C6H15N
471283
Triethylamine, ≥99.5%
C6H15N
W424601
Triethylamine, ≥99.5%
C6H15N