EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Instability-Assisted Direct Writing of Microstructured Fibers Featuring Sacrificial Bonds.
PMID 25981207
Abstract
A 30 μm-diameter thread of poly(lactic acid) (PLA) dissolved in dichloromethane is deposited on top of the eye of a sewing needle. The deposition robot traces a straight line; the helical shape of the thread is due to the liquid rope coiling instability. This instability is used to fabricate microstructured fibers with tailored mechanical properties.