EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Transition of serine residues to the D-form during the conversion of ovalbumin into heat stable S-ovalbumin.
PMID 25982752
Abstract
Ovalbumin, a major protein in chicken egg white, is converted into a more thermostable molecular form, known as S-ovalbumin, during the storage of shell eggs. Our previous X-ray crystallographic study indicated that S-ovalbumin contains three D-Ser residues (S164, S236, and S320), which may account for its thermostability. Here, we confirmed the presence of these D-Ser residues in ovalbumin using a technique combining deuterium labeling of α-protons of amino acids and liquid chromatography-tandem mass spectrometry (LC-MS/MS). Ovalbumin from chicken egg white and recombinant ovalbumin were incubated for approximately 12 days at pH 9.5 and 37°C. They were then hydrolyzed in DCl/D2O vapor, derivatized with 4-fluoro-7-nitro-2,1,3-benzoxadiazole (NBD-F), and analyzed by LC-MS/MS. A time-dependent increase in the D-Ser contents in native ovalbumin was observed over a period of 7 days, reaching approximately 8%. This corresponds to a value of three serine residues per molecule, and is consistent with the prediction based on our previous crystallographic analysis. Nearly identical results were obtained with recombinant ovalbumin. We then used this technique to investigate whether D-amino acid residues could arise within other proteins under mild alkaline conditions and detected small but significant amounts of D-Ala and/or D-Ser residues that increased in a time-dependent manner in some proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7641 Albumin from chicken egg white, lyophilized powder
488682
Deuterium chloride, 99 atom % D
DCl
543047
Deuterium chloride solution, 35 wt. % in D2O, ≥99 atom % D
DCl
532991
Deuterium chloride solution, 1.0 M in diethyl ether, 97.5 atom % D
DCl
633178
Deuterium oxide, 99.994 atom % D, contains 1 mM terephthalic acid disodium salt, 0.01 % (w/v) DSS-d6
D2O
613398
Deuterium oxide, extra, 99.994 atom % D
D2O
191701
Deuterium oxide, "100%", 99.990 atom % D
D2O
364312
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
D2O
151890
Deuterium oxide, "100%", ≥99.96 atom % D
D2O
269786
Deuterium oxide, "100%", 99.96 atom % D
D2O
151882
Deuterium oxide, 99.9 atom % D
D2O
530867
Deuterium oxide, 99.9 atom % D
D2O
613444
Deuterium oxide, 99.9 atom % D, glass distilled
D2O
450510
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
D2O
293040
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
D2O
343773
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
D2O
347167
Deuterium oxide, 99.9 atom % D, ~150 dpm/mL tritium
D2O
617385
Deuterium oxide, 99.8 atom % D
D2O
435767
Deuterium oxide, 99 atom % D
D2O
613436
Deuterium oxide, 60 atom % D
D2O
613428
Deuterium oxide, 70 atom % D
D2O
661643
Deuterium oxide, 99.99 atom % D (for D2O), contains 1% DSS-d6
D2O
756822
Deuterium oxide, filtered, 99.8 atom % D
D2O
P5629
Potassium phosphate tribasic, reagent grade, ≥98%
K3O4P