EMAIL THIS PAGE TO A FRIEND
Experimental cell research
Secretome protein signature of human gastrointestinal stromal tumor cells.
PMID 25983130
Abstract
Strategies for correct diagnosis, treatment evaluation and recurrence prediction are important for the prognosis and mortality rates among cancer patients. In spite of major improvements in clinical management, gastrointestinal stromal tumors (GISTs) can still be deadly due to metastasis and recurrences, which confirms the unmet need of reliable follow-up modalities. Tumor-specific secreted, shed or leaked proteins (collectively known as secretome) are considered promising sources for biomarkers, and suitable for detection in biofluids. Herein, we stimulated cell secretion in the imatinib-sensitive GIST882 cell line and profiled the secretome, collected as conditioned media, by using a shotgun proteomics approach. We identified 764 proteins from all conditions combined, 51.3% being predicted as classically/non-classically secreted. The protein subsets found were dependent on the stimulatory condition. The significant increase in protein release by the classical pathway was strongly associated with markers already found in other cancer types. Furthermore, most of the released proteins were non-classically released and overlapped to a high degree with proteins of exosomal origin. Imatinib pre-treatment radically changed these secretory patterns, which can have clinical implications when investigating biomarkers in imatinib-treated versus non-treated GIST patients. Our results show, for the first time, that GISTs contain a secretome signature. In the search for suitable biomarkers in the more complex GIST patient samples, this study aids in the understanding of basic GIST secretome characteristics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1306362
ANTI-HSP27 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501456
Anti-HSP27 antibody produced in rabbit, affinity isolated antibody
SAB4501457
Anti-HSP27 antibody produced in rabbit, affinity isolated antibody
SAB4501464
Anti-HSP60, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300495
Anti-HSPB1 (Ab-78) antibody produced in rabbit, affinity isolated antibody
SAB4300496
Anti-HSPB1 (Ab-82) antibody produced in rabbit, affinity isolated antibody
AV09060
Anti-HSPB1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV30177
Anti-HSPB1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA000497
Anti-HSPB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701965
Anti-HSPB1 antibody produced in rabbit
HPA038889
Anti-HSPB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB5200099 Anti-HSPB2 antibody produced in rabbit, 1 mg/mL, affinity isolated antibody
SAB4501458
Anti-HSPB2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1405973
Anti-HSPD1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001523
Anti-HSPD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107604
Anti-HSPD1 antibody produced in rabbit, affinity isolated antibody
SAB1410668
Anti-HSPD1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501909
Anti-MAT1 antibody produced in rabbit, affinity isolated antibody
SAB4503451
Anti-PEA-15 antibody produced in rabbit, affinity isolated antibody
SAB1306536
ANTI-PEA-15(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503450
Anti-PEA-15, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV48232
Anti-PEA15 antibody produced in rabbit, affinity isolated antibody
SAB4503452
Anti-PEA15 antibody produced in rabbit, affinity isolated antibody
H3524 Monoclonal Anti-Heat Shock Protein 60 antibody produced in mouse, clone LK2, ascites fluid
H4149 Monoclonal Anti-Heat Shock Protein 60 antibody produced in mouse, clone LK1, ascites fluid
SAB3701437
Monoclonal Anti-HSP27 antibody produced in mouse, clone G3.1, purified immunoglobulin
SAB4100596
Monoclonal Anti-HSPD1 antibody produced in mouse, clone HSPD1-A580, culture supernatant
SAB4100597
Monoclonal Anti-HSPD1 antibody produced in mouse, clone HSPD1-B580, culture supernatant
PLA0269
Rabbit anti-HSP60 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.