EMAIL THIS PAGE TO A FRIEND
BioMed research international
A Comprehensive Systems Biological Study of Autophagy-Apoptosis Crosstalk during Endoplasmic Reticulum Stress.
PMID 25984530
Abstract
One of the most important tasks of a living organism is to maintain its genetic integrity with respect to stress. Endoplasmic reticulum (ER) has a crucial role in sensing cellular homeostasis by controlling metabolism, proteostasis, and several signaling processes. ER stressors can induce autophagy-dependent survival; however excessive level of stress results in apoptotic cell death. Although many molecular components of these networks have already been discovered, the analysis of the dynamical features of the regulatory network of life-or-death decision is still lacking. Our goal was to incorporate both theoretical and molecular biological techniques to explore the autophagy-apoptosis crosstalk under ER stress. Using various levels of different ER stressors we confirmed that the control network always generated an evidently detectable autophagy-dependent threshold for apoptosis activation. We explored the features of this threshold by introducing both autophagy activators and inhibitors, and transient treatment with excessive level of ER stressor was also performed. Our experimental data were also supported by a stochastic approach. Our analysis suggests that even if the switch-like characteristic of apoptosis activation is hardly seen on population level the double negative feedback loop between autophagy and apoptosis inducers introduces bistability in the control network.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ROANNV
Annexin-V-FLUOS Staining Kit
SAB4503294
Anti-CASP3 (p17, Cleaved-Asp175) antibody produced in rabbit, affinity isolated antibody
SAB2501669
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002643
Anti-CASP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV00021
Anti-CASP3 antibody produced in rabbit, IgG fraction of antiserum
SAB4503291
Anti-CASP3 antibody produced in rabbit, affinity isolated antibody
SAB2701263
Anti-DCTN4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107220
Anti-DCTN4 antibody produced in rabbit, affinity isolated antibody
SAB2105017
Anti-DCTN4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW22005A Anti-GTF2H1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22005B Anti-GTF2H1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV100811
Anti-GTF2H1 antibody produced in rabbit, IgG fraction of antiserum
SAB2104463
Anti-GTF2H1 antibody produced in rabbit, affinity isolated antibody
L7543
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4200361
Anti-LC3B antibody, Mouse monoclonal, clone LC3B-6, purified from hybridoma cell culture
SAB2700738
Anti-Map1lc3b antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3501065
Anti-MAP1LC3B antibody produced in rabbit, affinity isolated antibody
GW22431A Anti-NUP62 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA005435
Anti-NUP62 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101664
Anti-NUP62 antibody produced in rabbit, affinity isolated antibody
SAB1410438
Anti-NUP62 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4500487
Anti-PARP (Cleaved-Asp214) antibody produced in rabbit, affinity isolated antibody
SAB4500488
Anti-PARP (Cleaved-Gly215) antibody produced in rabbit, affinity isolated antibody
P7605 Anti-PARP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV33754
Anti-PARP1 antibody produced in rabbit, IgG fraction of antiserum
HPA045168
Anti-PARP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000836M2 Monoclonal Anti-CASP3 antibody produced in mouse, clone 4D3, purified immunoglobulin, buffered aqueous solution
C5737 Monoclonal Anti-Caspase 3 antibody produced in mouse, clone 84803.111, purified immunoglobulin, lyophilized powder
WH0002965M1
Monoclonal Anti-GTF2H1 antibody produced in mouse, clone 1F12-1B5, purified immunoglobulin, buffered aqueous solution
SAB1403894
Monoclonal Anti-GTF2H1, (C-terminal) antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution