EMAIL THIS PAGE TO A FRIEND
Nature methods
Probing a cell-embedded megadalton protein complex by DNP-supported solid-state NMR.
PMID 25984698
Abstract
Studying biomolecules at atomic resolution in their native environment is the ultimate aim of structural biology. We investigated the bacterial type IV secretion system core complex (T4SScc) by cellular dynamic nuclear polarization-based solid-state nuclear magnetic resonance spectroscopy to validate a structural model previously generated by combining in vitro and in silico data. Our results indicate that T4SScc is well folded in the cellular setting, revealing protein regions that had been elusive when studied in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D198501
2,4-Dinitrophenol, moistened with water, ≥98.0%
C6H4N2O5