EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Siglec-G Deficiency Leads to Autoimmunity in Aging C57BL/6 Mice.
PMID 25987743
Abstract
Siglec-G, a member of the sialic acid-binding Ig-like lectin (Siglec) family, is expressed on B cell and dendritic cell surfaces. It acts as an inhibitory coreceptor and modulates B cell activation, especially on B1 cells, as Siglec-G-deficient mice show mainly a B1 cell-restricted phenotype resulting in increased B1 cell numbers. Although higher B1 cell numbers are discussed to be associated with autoimmunity, loss of Siglec-G does not result in autoimmune disease in BALB/c mice. However, there is evidence from Siglec-G × CD22 double-deficient mice and Siglec-G(-/-) mice on an autoimmune-prone MRL/lpr background that Siglec-G is important to maintain tolerance in B cells. In this study, we analyzed the role of Siglec-G in induction and maintenance of B cell tolerance on C57BL/6 background and in the FcγRIIb-deficient background. We find that aging Siglec-G-deficient and Siglec-G × FcγRIIb double-deficient mice develop an autoimmune phenotype with elevated autoantibody levels and mild glomerulonephritis. Aging Siglec-G-deficient mice have elevated numbers of plasma cells and germinal center B cells, as well as a higher number of activated CD4 T cells, which likely all contribute to autoantibody production. Additional loss of the inhibitory receptor FcγRIIb in Siglec-G(-/-) mice does not result in exacerbation of disease. These results indicate that Siglec-G is important to maintain tolerance in B cells and prevent autoimmunity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2105133
Anti-CD5, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4503585
Anti-CD5, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA026872
Anti-FCER1G antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700389
Anti-FCER1G antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2106731
Anti-FCER1G antibody produced in rabbit, affinity isolated antibody
SAB1405799
Anti-FCER2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA008928
Anti-FCER2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408575
Anti-FCER2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA006185
Anti-SDC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1305542
MONOCLONAL ANTI-CD138 (C-TERMINAL) antibody produced in mouse, clone 587CT7.3.6.5, crude ascites, buffered aqueous solution
SAB1305543 MONOCLONAL ANTI-CD138 antibody produced in mouse, clone 480CT5.4.3, IgG fraction of antiserum, buffered aqueous solution
SAB1305487
MONOCLONAL ANTI-CD138 antibody produced in mouse, clone 480CT5.4.3, crude ascites, buffered aqueous solution
SAB4700245
Monoclonal Anti-CD21 antibody produced in mouse, clone LT21, purified immunoglobulin, buffered aqueous solution
SAB4700732 Monoclonal Anti-CD23 antibody produced in mouse, clone EBVCS-5, purified immunoglobulin, buffered aqueous solution
SAB4100025
Monoclonal Anti-CD5 antibody produced in mouse, culture supernatant
SAB4700066 Monoclonal Anti-CD5 antibody produced in mouse, clone CRIS1, purified immunoglobulin, buffered aqueous solution
SAB4700071 Monoclonal Anti-CD5 antibody produced in mouse, clone MEM-32, purified immunoglobulin, buffered aqueous solution
SAB4700730 Monoclonal Anti-CD5 antibody produced in mouse, clone L17F12, purified immunoglobulin, buffered aqueous solution
SAB1403808
Monoclonal Anti-FCER2 antibody produced in mouse, clone 2A7, purified immunoglobulin, buffered aqueous solution
SAB4100595
Monoclonal Anti-FCER2 antibody produced in mouse, clone FCER2-A577, culture supernatant
SAB1408993
Monoclonal Anti-FCER2 antibody produced in mouse, clone S51, ascites fluid, solution