EMAIL THIS PAGE TO A FRIEND
World journal of gastroenterology
Phagocytosis (cannibalism) of apoptotic neutrophils by tumor cells in gastric micropapillary carcinomas.
PMID 25987778
Abstract
To identify those with a micropapillary pattern, ascertain relative frequency and document clinicopathological characteristics by reviewing gastric carcinomas. One hundred and fifty-one patients diagnosed with gastric cancer who underwent gastrectomy were retrospectively studied and the presence of a regional invasive micropapillary component was evaluated by light microscopy. All available hematoxylin-eosin (HE)-stained slides were histologically reviewed and 5 tumors were selected as putative micropapillary carcinoma when cancer cell clusters without a vascular core within empty lymphatic-like space comprised at least 5% of the tumor. Tumor tissues from these 5 invasive gastric carcinomas were immunostained using an anti-mucin 1 (MUC1) antibody (clone MA695) to detect the characteristic inside-out pattern and with D2-40 antibody to determine the presence of intratumoral lymph vessels. Detection of intraepithelial neutrophil apoptosis was evaluated in consecutive histological tissue sections by three independent methods, namely light microscopy with HE staining, the conventional terminal deoxynucleotidyl transferase-mediated dUTP-biotin nick end-labeling (TUNEL) method and immunohistochemistry for activated caspase-3 (clone C92-605). Among 151 gastric cancers resected for cure, 5 (3.3%) were adenocarcinomas with a micropapillary component. Four of the patients died of disease from 6 to 23 mo and one patient was alive with metastases at 9 mo. All patients had advanced-stage cancer (≥ pT2) and lymph node metastasis. Positive MUC1 immunostaining on the stroma-facing surface (inside-out pattern) of the carcinomatous cluster cells, together with negative immunostaining for D2-40 in the cells limiting lymphatic-like spaces, confirmed the true micropapillary pattern in these gastric neoplasms. In all five cases, several micropapillae were infiltrated by neutrophils. HE staining, TUNEL assay and immunostaining for caspase-3 demonstrated apoptotic neutrophils within cytoplasmic vacuoles of tumor cells. These data suggest phagocytosis (cannibalism) of apoptotic neutrophils by micropapillary tumor cells. Tumor cell cannibalism is usually found in aggressive tumors with anaplastic morphology. Our data extend these observations to gastric micropapillary carcinoma: a tumor histotype analogously characterized by aggressive behavior and poor prognosis. The results are of interest because they raise the intriguing possibility that neutrophil cannibalism by tumor cells may be one of the mechanisms favoring tumor growth in gastric micropapillary carcinomas. This is the first study showing phagocytosis (cannibalism) of apoptotic neutrophils by tumor cells in gastric micropapillary carcinomas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501931
Anti-CD227, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501932
Anti-CD227, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2101529
Anti-MUC1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101530
Anti-MUC1 (ab2) antibody produced in rabbit, affinity isolated antibody
AV41446
Anti-MUC1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2101531
Anti-MUC1 (ab3) antibody produced in rabbit, affinity isolated antibody
GW22681 Anti-MUC1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA004179
Anti-MUC1 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA007235
Anti-MUC1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA008855
Anti-MUC1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501933
Anti-MUC1, C-Terminal antibody produced in rabbit, affinity isolated antibody
WH0004582M1
Monoclonal Anti-MUC1 antibody produced in mouse, clone 3B9, purified immunoglobulin, buffered aqueous solution
SAB4200017
Monoclonal Anti-MUC1 antibody produced in mouse, ~1.0 mg/mL, clone 1D1, purified immunoglobulin, buffered aqueous solution
SAB1404092
Monoclonal Anti-MUC1 antibody produced in mouse, clone 1B7, purified immunoglobulin, buffered aqueous solution