PloS one
Langerhans cell homeostasis and activation is altered in hyperplastic human papillomavirus type 16 E7 expressing epidermis.
PMID 25992642
Abstract
It has previously been shown that expression of human papillomavirus type 16 (HPV) E7 in epidermis causes hyperplasia and chronic inflammation, characteristics of pre-malignant lesions. Importantly, E7-expressing epidermis is strongly immune suppressed and is not rejected when transplanted onto immune competent mice. Professional antigen presenting cells are considered essential for initiation of the adaptive immune response that results in graft rejection. Langerhans cells (LC) are the only antigen presenting cells located in normal epidermis and altered phenotype and function of these cells may contribute to the immune suppressive microenvironment. Here, we show that LC are atypically activated as a direct result of E7 expression in the epidermis, and independent of the presence of lymphocytes. The number of LC was significantly increased and the LC are functionally impaired, both in migration and in antigen uptake. However when the LC were extracted from K14E7 skin and matured in vitro they were functionally competent to present and cross-present antigen, and to activate T cells. The ability of the LC to present and cross-present antigen following maturation supports retention of full functional capacity when removed from the hyperplastic skin microenvironment. As such, opportunities are afforded for the development of therapies to restore normal LC function in hyperplastic skin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7149
Ammonium thiocyanate, powder
CH4N2S
GW22573F Anti-BB1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1407414
Anti-CD207 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA011216
Anti-CD207 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401669
Anti-CD207 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV41929
Anti-CD40 antibody produced in rabbit, affinity isolated antibody
HPA031567
Anti-CD40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031568
Anti-CD40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103449
Anti-CD40, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4503674
Anti-CD40, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2501656
Anti-CD80 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405584
Anti-CD80 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1400126
Anti-HLA-E antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1401182
Anti-HLA-E antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA031454
Anti-HLA-E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000958M1
Monoclonal Anti-CD40 antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
AMAB90905
Monoclonal Anti-CD40 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1673, purified immunoglobulin, buffered aqueous glycerol solution
WH0000941M1 Monoclonal Anti-CD80 antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1403647
Monoclonal Anti-CD80 antibody produced in mouse, clone 3E10, ascites fluid
SAB4700140 Monoclonal Anti-CD80 antibody produced in mouse, clone MEM-233, purified immunoglobulin, buffered aqueous solution
SAB4700300 Monoclonal Anti-HLA-E antibody produced in mouse, clone MEM-E/08, purified immunoglobulin, buffered aqueous solution
SAB4700301 Monoclonal Anti-HLA-E antibody produced in mouse, clone MEM-E/06, purified immunoglobulin, buffered aqueous solution
SAB4700302 Monoclonal Anti-HLA-E antibody produced in mouse, clone MEM-E/07, purified immunoglobulin, buffered aqueous solution