PloS one
Toddaculin, Isolated from of Toddalia asiatica (L.) Lam., Inhibited Osteoclastogenesis in RAW 264 Cells and Enhanced Osteoblastogenesis in MC3T3-E1 Cells.
PMID 25993011
Abstract
Osteoporosis with bone loss is widely recognized as a major health problem. Bone homeostasis is maintained by balancing bone formation and bone resorption. The imbalance caused by increased bone resorption over bone formation can lead to various bone-related diseases such as osteoporosis and rheumatoid arthritis. Osteoclasts are the principal cells responsible for bone resorption and the main targets of anti-resorptive therapies. However, excessive inhibition of osteoclast differentiation may lead to inhibition of osteoblast differentiation. Therefore, it is important to screen for new compounds capable of inhibiting bone resorption and enhancing bone formation. Toddalia asiatica (L.) Lam. has been utilized traditionally for medicinal purposes such as the treatment of rheumatism. Currently, the extract is considered to be a good source of pharmacological agents for the treatment of bone-related diseases, but the active compounds have yet to be identified. We investigated whether toddaculin, derived from Toddalia asiatica (L.) Lam., affects both processes by inhibiting bone resorption and enhancing bone formation. Towards this end, we used pre-osteoclastic RAW 264 cells and pre-osteoblastic MC3T3-E1 cells. We found that toddaculin not only inhibited the differentiation of osteoclasts via activation of the NF-κB, ERK 1/2, and p38 MAPK signaling pathways, but it also induced differentiation and mineralization of osteoblasts by regulating differentiation factors. Thus, toddaculin might be beneficial for the prevention and treatment of osteoporosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
J104
Anti-c-Jun antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4300307
Anti-JUN (Ab-170) antibody produced in rabbit, affinity isolated antibody
SAB4300308
Anti-JUN (Ab-239) antibody produced in rabbit, affinity isolated antibody
SAB4300309
Anti-JUN (Ab-243) antibody produced in rabbit, affinity isolated antibody
SAB4300286
Anti-JUN (Ab-63) antibody produced in rabbit, affinity isolated antibody
SAB4300288
Anti-JUN (Ab-73) antibody produced in rabbit, affinity isolated antibody
SAB4300305
Anti-JUN (Ab-91) antibody produced in rabbit, affinity isolated antibody
SAB4300306
Anti-JUN (Ab-93) antibody produced in rabbit, affinity isolated antibody
AV100614
Anti-JUN (AB1) antibody produced in rabbit, affinity isolated antibody
AV100708
Anti-JUN (AB2) antibody produced in rabbit, affinity isolated antibody
AV100911
Anti-JUN (AB3) antibody produced in rabbit, affinity isolated antibody
SAB1406055
Anti-JUN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV38133
Anti-JUN antibody produced in rabbit, IgG fraction of antiserum
SAB2103656
Anti-JUN antibody produced in rabbit, affinity isolated antibody
SAB4500490
Anti-p38 MAPK, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300200
Anti-phospho-MAPK14 (pThr180) antibody produced in rabbit, affinity isolated antibody
SAB4300201
Anti-phospho-MAPK14 (pTyr182) antibody produced in rabbit, affinity isolated antibody
SAB4504095
Anti-phospho-p38 MAPK (pThr179+Tyr181) antibody produced in rabbit, affinity isolated antibody
SAB4504497
Anti-phospho-p38 MAPK (pThr180) antibody produced in rabbit, affinity isolated antibody
SAB4504498
Anti-phospho-p38 MAPK (pTyr182) antibody produced in rabbit, affinity isolated antibody
SAB4504094
Anti-phospho-p38 MAPK (pTyr322) antibody produced in rabbit, affinity isolated antibody
SAB5300271
Monoclonal Anti-JUN antibody produced in mouse, clone 5B1, ascites fluid
SAB5300272
Monoclonal Anti-JUN antibody produced in mouse, clone 4H9, ascites fluid
SAB4100540
Monoclonal Anti-JUN antibody produced in mouse, clone JUN-A487, culture supernatant
SAB4100541
Monoclonal Anti-JUN antibody produced in mouse, clone JUN-B487, culture supernatant
SAB1403701
Monoclonal Anti-MAPK14, (C-terminal) antibody produced in mouse, clone 3D5, ascites fluid
PLA0273
Rabbit anti-c-Jun Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.