EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Synergistic Potentials of Coffee on Injured Pancreatic Islets and Insulin Action via KATP Channel Blocking in Zebrafish.
PMID 25993315
Abstract
Pancreatic islets (PIs) are damaged under diabetic conditions, resulting in decreased PI size. This study examined the regenerative effects of coffee and its components (caffeine, CFI; trigonelline, TRG; chlorogenic acid, CGA) on zebrafish larval PIs and β-cells damaged by administration of alloxan (AX). In addition, the influence of coffee and its active components on KATP channels was investigated using diazoxide (DZ) as a KATP channel activator. PI size and fluorescence intensity were significantly increased in the coffee-treated group relative to the no-treatment group (P < 0.0001). In addition, coffee exerted significant regenerative effects on pancreatic β-cells (p = 0.006). Treatment with TRG and CGA rescued PI damage, and the combination of TRG/CGA had a synergistic effect. In conclusion, the results indicate that coffee has beneficial effects on AX-damaged PIs and may also be useful as a blocker of pancreatic β-cell K(+) channels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9035
Diazoxide
C8H7ClN2O2S
G2295
Glimepiride, ≥98% (HPLC), solid
C24H34N4O5S