EMAIL THIS PAGE TO A FRIEND
Acta biochimica et biophysica Sinica
Over-expression of recombinant proteins with N-terminal His-tag via subcellular uneven distribution in Escherichia coli.
PMID 25994007
Abstract
Specific tags with defined amino acid residues are widely used to purify or probe target proteins. Interestingly, the tagging system occasionally results in an increase of the recombinant protein expression in vivo. Here, we systematically examined this phenomenon using a poly-histidine (His)-tag fused to N- or C-terminal region of green, red, and blue fluorescent proteins by quantification and uneven distribution in cytoplasm of Escherichia coli. This effect was further supported by the distinct over-expression of several unrelated proteins, such as esterase, neopullulanase, and chloramphenicol acetyltransferase, tagging with the same tag. These results suggest that a poly-His-tag placed at N-terminal region can induce over-expression of recombinant protein via subcellular uneven distribution in vivo.