EMAIL THIS PAGE TO A FRIEND
The British journal of nutrition
In vitro selenium accessibility in pet foods is affected by diet composition and type.
PMID 25994047
Abstract
Se bioavailability in commercial pet foods has been shown to be highly variable. The aim of the present study was to identify dietary factors associated with in vitro accessibility of Se (Se Aiv) in pet foods. Se Aiv is defined as the percentage of Se from the diet that is potentially available for absorption after in vitro digestion. Sixty-two diets (dog, n 52; cat, n 10) were in vitro enzymatically digested: fifty-four of them were commercially available (kibble, n 20; pellet, n 8; canned, n 17; raw meat, n 6; steamed meat, n 3) and eight were unprocessed (kibble, n 4; canned, n 4) from the same batch as the corresponding processed diets. The present investigation examined if Se Aiv was affected by diet type, dietary protein, methionine, cysteine, lysine and Se content, DM, organic matter and crude protein (CP) digestibility. Se Aiv differed significantly among diet types (P
Related Materials