EMAIL THIS PAGE TO A FRIEND
Fertility and sterility
Human chorionic gonadotropin stimulates spheroid attachment on fallopian tube epithelial cells through the mitogen-activated protein kinase pathway and down-regulation of olfactomedin-1.
PMID 25999259
Abstract
To study the effect of human chorionic gonadotropin (hCG) on olfactomedin-1 (Olfm1) expression and spheroid attachment in human fallopian tube epithelial cells in vitro. Experimental study. Reproductive biology laboratory. Healthy nonpregnant women. No patient interventions. Luteinizing hormone/chorionic gonadotropin receptor (LHCGR) and Olfm1 expression in fallopian tube epithelium cell line (OE-E6/E7 cells). OE-E6/E7 cells treated with hCG, U0126 extracellular signal-regulated kinase (ERK) inhibitor, or XAV939 Wnt/β-catenin inhibitor were analyzed by Western blotting, real-time polymerase chain reaction, and in vitro spheroid attachment assay. Human chorionic gonadotropin increased spheroid attachment on OE-E6/E7 cells through down-regulation of Olfm1 and activation of Wnt and mitogen-activated protein kinase (MAPK) signaling pathways. U0126 down-regulated both MAPK and Wnt/β-catenin signaling pathways and up-regulated Olfm1 expression. XAV939 down-regulated only the Wnt/β-catenin signaling pathway but up-regulated Olfm1 expression. Human chorionic gonadotropin activated both ERK and Wnt/β-catenin signaling pathways and enhanced spheroid attachment on fallopian tube epithelial cells through down-regulation of Olfm1 expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV40173
Anti-ACTB antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2100037
Anti-ACTB antibody produced in rabbit, affinity isolated antibody
GW23014 Anti-Actin, Cytoplasmic 1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1100676
Anti-AXIN2 (151-165) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500619
Anti-AXIN2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA042344
Anti-AXIN2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500350
Anti-β-Actin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4200176
Anti-JNK antibody, Mouse monoclonal, clone 1C2, purified from hybridoma cell culture
SAB4502398
Anti-JNK1 antibody produced in rabbit, affinity isolated antibody
SAB2700732
Anti-LHCGR (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501752
Anti-LSHR, C-Terminal antibody produced in rabbit, affinity isolated antibody
L6792 Anti-Luteinizing Hormone/Choriogonadotropin Receptor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
M7556 Anti-MAP Kinase 2 (ERK-2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL
GW22768 Anti-MAPK1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA030069
Anti-MAPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300499
Anti-MAPK14 (Ab-182) antibody produced in rabbit, affinity isolated antibody
SAB1405665
Anti-MAPK14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV06004
Anti-MAPK14 antibody produced in rabbit, affinity isolated antibody
SAB1406300
Anti-MAPK8 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2702026
Anti-OLFM1 (C-terminal) antibody produced in rabbit
SAB1300303
Anti-OLFM1 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA057444
Anti-OLFM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
M0800
Anti-p38 MAP Kinase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4500491
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500492
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500493
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500490
Anti-p38 MAPK, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300200
Anti-phospho-MAPK14 (pThr180) antibody produced in rabbit, affinity isolated antibody
SAB4300201
Anti-phospho-MAPK14 (pTyr182) antibody produced in rabbit, affinity isolated antibody
P1491 Anti-phospho-p38 (pThr180/pTyr182) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution