EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Modulation of LSD1 phosphorylation by CK2/WIP1 regulates RNF168-dependent 53BP1 recruitment in response to DNA damage.
PMID 25999347
Abstract
Proper DNA damage response is essential for the maintenance of genome integrity. The E3 ligase RNF168 deficiency fully prevents both the initial recruitment and retention of 53BP1 at sites of DNA damage. In response to DNA damage, RNF168-dependent recruitment of the lysine-specific demethylase LSD1 to the site of DNA damage promotes local H3K4me2 demethylation and ubiquitination of H2A/H2AX, facilitating 53BP1 recruitment to sites of DNA damage. Alternatively, RNF168-mediated K63-linked ubiquitylation of 53BP1 is required for the initial recruitment of 53BP1 to sites of DNA damage and for its function in repair. We demonstrated here that phosphorylation and dephosphorylation of LSD1 at S131 and S137 was mediated by casein kinase 2 (CK2) and wild-type p53-induced phosphatase 1 (WIP1), respectively. LSD1, RNF168 and 53BP1 interacted with each other directly. CK2-mediated phosphorylation of LSD1 exhibited no impact on its interaction with 53BP1, but promoted its interaction with RNF168 and RNF168-dependent 53BP1 ubiquitination and subsequent recruitment to the DNA damage sites. Furthermore, overexpression of phosphorylation-defective mutants failed to restore LSD1 depletion-induced cellular sensitivity to DNA damage. Taken together, our results suggest that LSD1 phosphorylation modulated by CK2/WIP1 regulates RNF168-dependent 53BP1 recruitment directly in response to DNA damage and cellular sensitivity to DNA damaging agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-1130
Anti-RNF168 Antibody, 100 µg, Anti-RNF168 Antibody is a Rabbit Polyclonal Antibody for detection of RNF168 also known as E3 ubiquitin-protein ligase RNF168 & has been validated in WB.
B4436
Anti-53BP1 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
B4561
Anti-53BP1 (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1407168
Anti-AOF2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV53595
Anti-CSNK2A2 (AB1) antibody produced in rabbit, affinity isolated antibody
SAB1410253
Anti-CSNK2A2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306220 ANTI-LSD1 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB3500229
Anti-LSD1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500953
Anti-LSD1 antibody produced in rabbit, affinity isolated antibody
SAB1306581
ANTI-LSD1(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306580
ANTI-LSD1(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2101614
Anti-NOL3 antibody produced in rabbit, affinity isolated antibody
PRS2225
Anti-NOP30 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306802
ANTI-PPM1D (CENTER) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA022277
Anti-PPM1D antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306924 ANTI-PPM1D(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1408409
Anti-RNF168 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV43433
Anti-RNF168 antibody produced in rabbit, affinity isolated antibody
SAB3500207
Anti-RNF168 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1410293
Anti-RNF168 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA008788
Anti-TP53BP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA022133
Anti-TP53BP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
E1383
Etoposide, synthetic, ≥98%, powder
C29H32O13
PLA0303
Goat anti-53BP1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
WH0001459M1 Monoclonal Anti-CSNK2A2 antibody produced in mouse, clone 4F2, purified immunoglobulin, buffered aqueous solution
SAB1403706
Monoclonal Anti-CSNK2A2, (N-terminal) antibody produced in mouse, clone 1E8, purified immunoglobulin, buffered aqueous solution
L7293
Monoclonal Anti-LSD1 (AOF2) antibody produced in mouse, ~2 mg/mL, clone LSD1-12, purified immunoglobulin, buffered aqueous solution
WH0008493M1
Monoclonal Anti-PPM1D antibody produced in mouse, clone 4D1, purified immunoglobulin, buffered aqueous solution
WH0165918M1
Monoclonal Anti-RNF168 antibody produced in mouse, clone 3E1, purified immunoglobulin, buffered aqueous solution
WH0007158M1
Monoclonal Anti-TP53BP1 antibody produced in mouse, clone 1B9, purified immunoglobulin, buffered aqueous solution