EMAIL THIS PAGE TO A FRIEND
Journal of anesthesia
Sevoflurane suppresses hypoxia-induced growth and metastasis of lung cancer cells via inhibiting hypoxia-inducible factor-1α.
PMID 26002230
Abstract
Hypoxia promotes the progression of lung cancer cells. Unfortunately, anesthetic technique might aggravate hypoxia of lung cancer cells. Sevoflurane is a commonly used anesthetic. Its effect on hypoxia-induced aggressiveness of lung cancer cells remains unknown. The aim of the study is to investigate the effects of sevoflurane on hypoxia-induced growth and metastasis of lung cancer cells. As hypoxia-inducible factor-1α (HIF-1α) plays a pivotal role in mediating the adaptation and tolerance of cancer cells under hypoxic microenvironment, the role of HIF-1α in the effect of sevoflurane on hypoxia-induced growth and metastasis has also been elucidated. A549 cells were treated with normoxia, hypoxia, co-treatment of sevoflurane and hypoxia, and dimethyloxaloylglycine (DMOG, a HIF-1α agonist) for 4xa0h, respectively. MTT assay and colony formation assay were used to evaluate cell growth. Transwell assay was performed to detect invasion and migration ability. The protein level of HIF-1α, X-linked inhibitor of apoptosis protein (XIAP), survivin, fascin, heparanase (HPA), and p38 MAPK were determined by Western blotting. Hypoxia enhanced proliferation and metastatic potential of cells. Sevoflurane could suppress hypoxia-induced growth and metastasis ability of cells. Furthermore, HIF-1α, XIAP, survivin, fascin and HPA were down-regulated significantly by the co-treatment of sevoflurane and hypoxia as compared to hypoxia treatment. DMOG abolished the inhibiting effects of sevoflurane on hypoxia-induced growth and metastasis ability of cells. In addition, sevoflurane partly reversed the increase of p38 MAPK activity that was induced by hypoxia. Sevoflurane could suppress hypoxia-induced growth and metastasis of lung cancer cells, which might be associated with modulating HIF-1α and its down-stream genes. Moreover, p38 MAPK signaling pathway was involved in the regulation of HIF-1α by sevoflurane.