EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Amido-Schiff base derivatives as colorimetric fluoride sensor: Effect of nitro substitution on the sensitivity and color change.
PMID 26002437
Abstract
A series of Schiff bases synthesized by the condensation of benzohydrazide and -NO2 substituted benzaldehyde have been used as selective fluoride ion sensor. Test paper coated with these synthetic Schiff bases (test kits) can detect fluoride ion selectively with a drastic color change and detection can be achieved by just using the naked-eye without the help of any optical instrument. Interestingly, the position of -NO2 group in the amido Schiff bases has an effect on the sensitivity as well as on the change of color of species.