EMAIL THIS PAGE TO A FRIEND
Laboratory investigation; a journal of technical methods and pathology
Bronchial lesions of mouse model of asthma are preceded by immune complex vasculitis and induced bronchial associated lymphoid tissue (iBALT).
PMID 26006019
Abstract
We systematically examined by immune histology the lungs of some widely used mouse models of asthma. These models include sensitization by multiple intraperitoneal injections of soluble ovalbumin (OVA) or of OVA with alum, followed by three intranasal or aerosol challenges 3 days apart. Within 24 h after a single challenge there is fibrinoid necrosis of arterial walls with deposition of immunoglobulin (Ig) and OVA and infiltration of eosinophilic polymorphonuclear cells that lasts for about 3 days followed by peribronchial B-cell infiltration and slight reversible goblet cell hypertrophy (GCHT). After two challenges, severe eosinophilic vasculitis is present at 6 h, increases by 72 h, and then declines; B-cell proliferation and significant GCHT and hyperplasia (GCHTH) and bronchial smooth muscle hypertrophy recur more prominently. After three challenges, there is significantly increased induced bronchus-associated lymphoid tissue (iBALT) formation, GCHTH, and smooth muscle hypertrophy. Elevated levels of Th2 cytokines, IL-4, IL-5, and IL-13, are present in bronchial lavage fluids. Sensitized mice have precipitating antibody and positive Arthus skin reactions but also develop significant levels of IgE antibody to OVA but only 1 week after challenge. We conclude that the asthma like lung lesions induced in these models is preceded by immune complex-mediated eosinophilic vasculitis and iBALT formation. There are elevations of Th2 cytokines that most likely produce bronchial lesions that resemble human asthma. However, it is unlikely that mast cell-activated atopic mechanisms are responsible as we found only a few presumed mast cells by toluidine blue and metachromatic staining limited to the most proximal part of the main stem bronchus, and none in the remaining main stem bronchus or in the lung periphery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5503
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
M0945 Anti-Maspin antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1306340
ANTI-PAI2 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1406218
Anti-SERPINB2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA015480
Anti-SERPINB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104454
Anti-SERPINB2 antibody produced in rabbit, affinity isolated antibody
SAB1410870
Anti-SERPINB2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA020136
Anti-SERPINB5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
HPA019025
Anti-SERPINB5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA019132
Anti-SERPINB5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB4502191
Anti-SERPINB5 antibody produced in rabbit, affinity isolated antibody
HPA024200
Anti-SERPINB7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401435
Anti-SERPINB7 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4503460
Anti-SERPINB7 antibody produced in rabbit, affinity isolated antibody
SAB1400210
Anti-SERPINB8 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2500930
Anti-SERPINB9 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA030067
Anti-SERPINB9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502192
Anti-SERPINB9 antibody produced in rabbit, affinity isolated antibody
E2886 ExtrAvidin®−Peroxidase, buffered aqueous solution
WH0005055M8
Monoclonal Anti-SERPINB2 antibody produced in mouse, clone 3A9, purified immunoglobulin, buffered aqueous solution
SAB1402869
Monoclonal Anti-SERPINB8 antibody produced in mouse, clone 1D10, ascites fluid
SAB1402309
Monoclonal Anti-SERPINB9, (C-terminal) antibody produced in mouse, clone 1F5, purified immunoglobulin, buffered aqueous solution