EMAIL THIS PAGE TO A FRIEND
Journal of cancer research and clinical oncology
Epigenetic priming of non-small cell lung cancer cell lines to the antiproliferative and differentiating effects of all-trans retinoic acid.
PMID 26008188
Abstract
The retinoic acid signaling pathway, crucial for differentiation, is silenced by epigenetic mechanisms in many cancers. Epigenetically active, chromatin-modifying agents offer a novel treatment approach, by reactivating aberrantly silenced genes in tumor cells and by sensitizing them to subsequent treatments. We hypothesized that the treatment of non-small cell lung cancer (NSCLC) cells with a histone deacetylase (HDAC) inhibitor may prime them to the antiproliferative and differentiating activity of all-trans retinoic acid. The NSCLC cell lines A549, NCI-H460 and HCC827 were treated with ATRA (2 µM) and the pan-HDAC inhibitor panobinostat (LBH589; 10-35 nM). While treatment with ATRA alone showed only very modest effects, panobinostat reduced cellular proliferation by at least 50 %. Notably, the combination of panobinostat and ATRA had additive and synergistic effects, respectively, on growth inhibition and differentiation, with almost no cytotoxicity. Effects were strongest in A549, followed by the EGFR-mutant HCC827, and least pronounced in NCI-H460. Global histone H3 acetylation was strongly induced by panobinostat; interestingly, ATRA alone had also an effect on histone acetylation, which was synergistically enhanced when the HDAC inhibitor was added. The combination of the two drugs additively decreased expression of phospho-ERK and phospho-AKT, whereas p53 and p21(CIP1/WAF1) proteins were both induced. Panobinostat sensitized, to varying degrees, all three cell lines to the antiproliferative and differentiating effects of ATRA, with synergistic histone H3 acetylation. Combination therapy with an epigenetic drug and ATRA may offer an alternative to aggressive chemotherapy even in primary ATRA-insensitive tumors, such as adenocarcinomas of the lung.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
364959
N-[Tris(hydroxymethyl)methyl]acrylamide, contains ≤7% KCl, 93%
C7H13NO4