PloS one
CRMP1 Inhibits Proliferation of Medulloblastoma and Is Regulated by HMGA1.
PMID 26009886
Abstract
Many facets of the tumor biology of medulloblastoma (MB) have not been fully elucidated. Collapsin response mediator protein 1 (CRMP1) is a member of cytoplasmic family of proteins that regulate the development of central nervous system. Recent studies demonstrated that CRMP1 could function as an invasion suppressor. We reported previously that high mobility group AT-hook 1 (HMGA1) contributed to development of MB and regulated its growth and migration/invasion. Transcriptional profiling and quantitative RT-PCR revealed increased expression of CRMP1 in HMGA1-depleted cells, suggesting that CRMP1 may be a downstream target of HMGA1 in MB. In this study, we showed HMGA1 can bind CRMP1 promoter by chromatin immunoprecipitation (ChIP) assay. Luciferase assay demonstrated a marked enhancement of CRMP1 transcription activity in HMGA1-depleted cells. Furthermore, quantitative RT-PCR revealed a negative correlation between HMGA1 and CRMP1 in 32 MB samples. To investigate the biological roles of CRMP1 in MB pathogenesis, we established MB clones stably expressing CRMP1. Functional analysis revealed that expression of CRMP1 significantly inhibited proliferation, migration, invasion and formation of filopodia and intense stress fiber of MB cells. Our data suggest that HMGA1 regulates CRMP1 expression and CRMP1 is implicated in MB pathogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV41932
Anti-CRMP1 (AB1) antibody produced in rabbit, affinity isolated antibody
AV41934
Anti-CRMP1 (AB2) antibody produced in rabbit, affinity isolated antibody
C4118
Anti-CRMP1 (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
C2868
Anti-CRMP1 antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2100485
Anti-CRMP1 antibody produced in rabbit, affinity isolated antibody
SAB3500201
Anti-CRMP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA035640
Anti-CRMP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501157 Anti-HMGA1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501305
Anti-HMGA1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1401183
Anti-HMGA1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1409197
Anti-HMGA1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1402156
Monoclonal Anti-CRMP1 antibody produced in mouse, clone 2B6, purified immunoglobulin, buffered aqueous solution
SAB1402806
Monoclonal Anti-HMGA1 antibody produced in mouse, clone 2A1, purified immunoglobulin, buffered aqueous solution