EMAIL THIS PAGE TO A FRIEND
Oncotarget
Phosphorylation of interleukin (IL)-24 is required for mediating its anti-cancer activity.
PMID 26009991
Abstract
Interleukin (IL)-24 is a tumor suppressor/cytokine gene that undergoes post-translational modifications (PTMs). Glycosylation and ubiquitination are important for IL-24 protein stabilization and degradation respectively. Little is known about IL-24 protein phosphorylation and its role in IL-24-mediated anti-tumor activities. In this study we conducted molecular studies to determine whether IL-24 phosphorylation is important for IL-24-mediated anti-cancer activity.Human H1299 lung tumor cell line that was stably transfected with a doxycycline (DOX)-inducible (Tet-on) plasmid vector carrying the cDNA of IL-24-wild-type (IL-24wt) or IL-24 with all five phosphorylation sites replaced (IL-24mt) was used in the present study. Inhibition of tumor cell proliferation, cell migration and invasion, and induction of G2/M cell cycle arrest was observed in DOX-induced IL-24wt-expressing cells but not in IL-24mt-expressing cells. Secretion of IL-24mt protein was greatly reduced compared to IL-24wt protein. Further, IL-24wt and IL-24mt proteins markedly differed in their subcellular organelle localization. IL-24wt but not IL-24mt inhibited the AKT/mTOR signaling pathway. SiRNA-mediated AKT knockdown and overexpression of myristolyated AKT protein confirmed that IL-24wt but not IL-24mt mediated its anti-cancer activity by inhibiting the AKT signaling pathway.Our results demonstrate that IL-24 phosphorylation is required for inhibiting the AKT/mTOR signaling pathway and exerting its anti-cancer activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1407085
Anti-IL24 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1408664
Anti-IL24 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1304456
ANTI-PDIA2 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2501845
Anti-PDIA2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501851
Anti-PDIA2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA051692
Anti-PDIA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA053492
Anti-PDIA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405897
Anti-PDIA3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002645
Anti-PDIA3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA003230
Anti-PDIA3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107799
Anti-PDIA3 antibody produced in rabbit, affinity isolated antibody
SAB1410582
Anti-PDIA3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
RAB0698 Human IL24 / Interleukin-24 ELISA Kit
EHU083501
MISSION® esiRNA, esiRNA human AKT1 (esiRNA1)
EMU015011 MISSION® esiRNA, esiRNA targeting mouse Akt1 (esiRNA1)
WH0011009M1
Monoclonal Anti-IL24 antibody produced in mouse, clone 4B6, purified immunoglobulin, buffered aqueous solution
P0878
O-Phospho-L-serine
C3H8NO6P
P1053
O-Phospho-L-threonine
C4H10NO6P